Skip to Content
Merck
All Photos(4)

Documents

S7388

Sigma-Aldrich

N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide

Synonym(s):

N-Succinyl-L-alanyl-L-alanyl-L-prolyl-L-phenylalanine 4-nitroanilide, N-Succinyl-Ala-Ala-Pro-Phe-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H36N6O9
CAS Number:
Molecular Weight:
624.64
Beilstein:
4289896
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

storage temp.

−20°C

SMILES string

[H][C@]1(CCCN1C(=O)[C@H](C)NC(=O)[C@H](C)NC(=O)CCC(O)=O)C(=O)N[C@@H](Cc2ccccc2)C(=O)Nc3ccc(cc3)[N+]([O-])=O

InChI

1S/C30H36N6O9/c1-18(31-25(37)14-15-26(38)39)27(40)32-19(2)30(43)35-16-6-9-24(35)29(42)34-23(17-20-7-4-3-5-8-20)28(41)33-21-10-12-22(13-11-21)36(44)45/h3-5,7-8,10-13,18-19,23-24H,6,9,14-17H2,1-2H3,(H,31,37)(H,32,40)(H,33,41)(H,34,42)(H,38,39)/t18-,19-,23-,24-/m0/s1

InChI key

LKDMKWNDBAVNQZ-WJNSRDFLSA-N

Gene Information

human ... FKBP10(60681)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

N-Suc-Ala-Ala-Pro-Phe-pNA

Application

N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide has been used as a substrate to determine chymotrypsin activity.
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide has been used:
  • as a substrate for cathepsin G assay in bone marrow lysates.
  • in proteolytic activity assay of serine protease subtilisin carlsberg (SC).
  • as a substrate for neutrophil elastase, trypsin and chymotrypsin in protease assay.

Biochem/physiol Actions

N-succinyl-Ala-Ala-Pro-Phe p-nitroanilide is a substrate for subtilisin and proteases.

Substrates

N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide is a colorimetric substrate for chymotrypsin, human leukocyte cathepsin G and peptidyl prolyl isomerase.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved Enantioselectivity of Subtilisin Carlsberg towards Secondary Alcohols by Protein Engineering
Dorau R, et al.
Chembiochem, 19(4), 338-346 (2018)
J J Siekierka et al.
Nature, 341(6244), 755-757 (1989-10-26)
CYCLOSPORIN A and the newly discovered immunosuppressant, FK-506, are potent inhibitors of T cell activation. In addition to their clinical importance in the prevention of allograft rejection, cyclosporin A and FK-506 represent important reagents for the study of the molecular
Jolanda Neef et al.
Microbial cell factories, 16(1), 57-57 (2017-04-06)
The microbial cell factory Bacillus subtilis is a popular industrial platform for high-level production of secreted technical enzymes. Nonetheless, the effective secretion of particular heterologous enzymes remains challenging. Over the past decades various studies have tackled this problem, and major
Interleukin-33 is activated by allergen-and necrosis-associated proteolytic activities to regulate its alarmin activity during epithelial damage
Scott IC, et al.
Scientific Reports, 8(1), 3363-3363 (2018)
Marco Bonelli et al.
International journal of molecular sciences, 21(14) (2020-07-17)
Modulation of nutrient digestion and absorption is one of the post-ingestion mechanisms that guarantees the best exploitation of food resources, even when they are nutritionally poor or unbalanced, and plays a pivotal role in generalist feeders, which experience an extreme

Articles

Analytical Enzyme Chymotrypsin: Chymotrypsin is produced in the acinar cells of the pancreas as the inactive precursor, chymotrypsinogen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service