Skip to Content
Merck
All Photos(2)

Key Documents

C9521

Sigma-Aldrich

Z-Phe-Arg 7-amido-4-methylcoumarin hydrochloride

kallikrein substrate, ≥95% (HPLC), powder

Synonym(s):

Z-Phe-Arg-AMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H36N6O6 · HCl
CAS Number:
Molecular Weight:
649.14
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Z-Phe-Arg 7-amido-4-methylcoumarin hydrochloride, kallikrein substrate

Assay

≥95% (HPLC)

form

powder

concentration

≥95%

solubility

methanol: 20 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

O=C(N[C@@H](CC1=CC=CC=C1)C(N[C@@H](CCCNC(N)=N)C(NC2=CC=C(C(C)=CC(O3)=O)C3=C2)=O)=O)OCC4=CC=CC=C4.[Cl]

InChI

1S/C33H36N6O6/c1-21-17-29(40)45-28-19-24(14-15-25(21)28)37-30(41)26(13-8-16-36-32(34)35)38-31(42)27(18-22-9-4-2-5-10-22)39-33(43)44-20-23-11-6-3-7-12-23/h2-7,9-12,14-15,17,19,26-27H,8,13,16,18,20H2,1H3,(H,37,41)(H,38,42)(H,39,43)(H4,34,35,36)

InChI key

ZZGDDBWFXDMARY-UHFFFAOYSA-N

General description

Z-Phe-Arg 7-amido-4-methylcoumarin (Z-FR-AMC) is a peptidomimetic substrate for papainand other enzymes such as cathepsin K. It is also a fluorogenic synthetic peptide for the enzymes cathepsins L and B.

Application

Z-Phe-Arg 7-amido-4-methylcoumarin hydrochloride has been used:
  • as a fluorogenic substrate in actinidin inhibition assay
  • as a kallikrein substrate
  • as a trypsin substrate for fluorometric assay
  • as a cathepsin-L substrate

Biochem/physiol Actions

Z-Phe-Arg 7-amido-4-methylcoumarin (Z-FR-AMC) proteolytic lysis by proteases leads to the liberation of AMC resulting in increased fluorescence in the enzymatic reaction.

Substrates

A fluorogenic substrate for plasma kallikrein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cinthia Bernardes Gomes et al.
Biochimie, 133, 28-36 (2016-12-07)
Leishmania (Viannia) braziliensis presents adaptive protease-dependent mechanisms, as cysteine proteinases B (CPB). This study investigates the expression of three cpb gene isoforms and CPB enzymatic activity during the parasite differentiation. Relative expression levels of LbrM.08.0810 gene were assessed, exhibiting a
Early ontogenetic development, digestive enzymatic activity and gene expression in red sea bream (Pagrus major)
Khoa TND, et al.
Aquaculture (Amsterdam, Netherlands), 512, 734283-734283 (2019)
Quantification of Functional Actinidin in Whole Kiwifruit Extract Using the Selective Cysteine Proteinase Inhibitor E-64
Martin H
Journal of Food & Nutrition Research, 4(4), 243-250 (2016)
R M C Deshapriya et al.
Journal of biochemistry, 147(3), 393-404 (2009-11-17)
To identify functionally essential sequences and residues of CTLA-2alpha, in vitro mutagenesis was carried out. The coefficient of inhibition (K(i)) was determined towards rabbit cathepsin L using Z-Phe-Arg-MCA as the substrate. Recombinant CTLA-2alpha inhibited the enzyme potently (K(i) = 15
H Ghoneim et al.
International journal for parasitology, 25(12), 1515-1519 (1995-12-01)
A previously described "major acidic proteinase" of adult Schistosoma mansoni, believed to play a key role in the parasite's metabolism, has been identified as a cathepsin B (Sm31). Purified Sm cathepsin B was not recognized by anti-Sm32 or anti-cathepsin L

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service