Skip to Content
Merck
All Photos(1)

Key Documents

S3652

Sigma-Aldrich

Shields and Sang M3 Insect Medium

With ʟ-glutamine and potassium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture

Synonym(s):

Insect Medium, M3 Insect Medium, Sang Insect Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

Shields and Sang M3 Insect Medium, With L-glutamine and potassium bicarbonate., liquid, sterile-filtered, suitable for insect cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | insect: suitable

components

glucose: 10 g/L (Dextro)
L-glutamine: 0.6 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Shields and Sang M3 Insect Medium has been used to culture Drosophila cell lines, larvae, and testes. 
Developed for the culture of Drosophila cell lines.

Biochem/physiol Actions

Insect tissue culture media have been formulated to mimic the main properties of the hemolymph of specific insects. A survey of the formulas of these media reveals significant differences in their composition. When supplemented with 5-20% heat-inactivated fetal bovine serum, Shields and Sang M3 medium has been found to support the rapid growth of both primary and established cultures of Drosophila melanogaster cells.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giri Raj Dahal et al.
Development (Cambridge, England), 144(15), 2771-2783 (2017-07-08)
Loss of embryonic ion channel function leads to morphological defects, but the underlying reason for these defects remains elusive. Here, we show that inwardly rectifying potassium (Irk) channels regulate release of the
Rosalind Hale et al.
eLife, 4 (2015-02-25)
To understand how long-range patterning gradients are interpreted at the cellular level, we investigate how a gradient of expression of the Four-jointed kinase specifies planar polarised distributions of the cadherins Fat and Dachsous in the Drosophila wing. We use computational
Remigio Picone et al.
PLoS biology, 8(11), e1000542-e1000542 (2010-11-26)
Because physical form and function are intimately linked, mechanisms that maintain cell shape and size within strict limits are likely to be important for a wide variety of biological processes. However, while intrinsic controls have been found to contribute to
Shuhei Kimura
FEBS letters, 587(7), 922-929 (2013-03-05)
Spermiogenesis is a dynamic process leading to alterations in cell morphology. In spermiogenesis, the roles of the histone chaperones are largely unknown. Here, I report the unexpected roles of two Nap family proteins, CG5017/Hanabi and nucleosome assembly protein 1 (Nap1)
Stephen K K Chan et al.
PLoS genetics, 13(11), e1007096-e1007096 (2017-11-21)
Repressors are frequently deployed to limit the transcriptional response to signalling pathways. For example, several co-repressors interact directly with the DNA-binding protein CSL and are proposed to keep target genes silenced in the absence of Notch activity. However, the scope

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service