Skip to Content
Merck
All Photos(1)

Documents

S0146

Sigma-Aldrich

Schneider′s Insect Medium

With ʟ-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture

Synonym(s):

Schneider’s Drosophila Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

Schneider′s Insect Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | insect: suitable

suitability

suitable for culture of other dipteran cell lines (Originally developed for the culture of Drosophila cells)

components

NaHCO3: 0.4 g/L
glucose: 2 g/L (Dextro)
L-glutamine: 1.8 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Schneider′s insect medium has been used:
  • to culture Drosophila neuronal cell line (BG3-c2 cells)
  • to cultivate subcultures of Leishmania major for promastigote growth inhibition assay
  • to seed purified ookinetes with Drosophila melanogaster S2 cells to assess the effect of the avermectin compound on cell development and to promote oocyte development

Originally developed for the culture of Drosophila; suitable for culture of other dipteran cell lines.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Franz et al.
The Journal of cell biology, 203(5), 785-799 (2013-12-04)
CP110 is a conserved centriole protein implicated in the regulation of cell division, centriole duplication, and centriole length and in the suppression of ciliogenesis. Surprisingly, we report that mutant flies lacking CP110 (CP110Δ) were viable and fertile and had no
Eva Wegel et al.
Scientific reports, 6, 27290-27290 (2016-06-07)
Many biological questions require fluorescence microscopy with a resolution beyond the diffraction limit of light. Super-resolution methods such as Structured Illumination Microscopy (SIM), STimulated Emission Depletion (STED) microscopy and Single Molecule Localisation Microscopy (SMLM) enable an increase in image resolution
Amit Sinha et al.
Genome biology and evolution, 11(3), 706-720 (2019-02-05)
Wolbachia, an alpha-proteobacterium closely related to Rickettsia, is a maternally transmitted, intracellular symbiont of arthropods and nematodes. Aedes albopictus mosquitoes are naturally infected with Wolbachia strains wAlbA and wAlbB. Cell line Aa23 established from Ae. albopictus embryos retains only wAlbB
Poonam Aggarwal et al.
Genetics, 207(1), 197-213 (2017-07-12)
One of the pertinent issues associated with cellular plasticity is to understand how the delicate balance between the determined state of cells and the extent to which they can transdetermine is maintained. Employing the well-established model of generating ectopic eyes
Amanda R Brock et al.
Genetics, 206(3), 1505-1520 (2017-05-18)
Regeneration is a complex process that requires an organism to recognize and repair tissue damage, as well as grow and pattern new tissue. Here, we describe a genetic screen to identify novel regulators of regeneration. We ablated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service