Skip to Content
Merck
All Photos(4)

Key Documents

237205

Sigma-Aldrich

Potassium bicarbonate

ACS reagent, 99.7%, powder, crystals or granules

Synonym(s):

Potassium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
Beilstein:
4535309
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
99.7%
99.7-100.5% (ACS specification)
grade:
ACS reagent
form:
powder, crystals or granules
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

99.7%
99.7-100.5% (ACS specification)

form

powder, crystals or granules

impurities

≤0.003% S compounds
≤0.01% insolubles

pH

8.2 (10.01 g/L)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤5 ppm

cation traces

Ca: ≤0.002%
Fe: ≤5 ppm
Mg: ≤0.001%
NH4+: ≤5 ppm
Na: ≤0.03%
heavy metals (as Pb): ≤5 ppm

SMILES string

[K+].OC([O-])=O

InChI

1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

TYJJADVDDVDEDZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium bicarbonate is a commercially available nontoxic base frequently used as a reagent as well as a pH regulator in research and industrial applications.

Application

Potassium bicarbonate can be used as:      
  • A catalyst to synthesize dimethyl carbonate and propylene glycol from propylene oxide, supercritical carbon dioxide, and methanol. In this process, KHCO3 also acts as a drying agent.      
  • A dopant in the epoxidation of propene in the presence of silica-supported vanadium oxide as a catalyst.      
  • A carboxyl source to synthesize Z-5-alkylidene-2-oxazolidinones via Ag2CO3 catalyzed carboxylative cyclization of propargylic amines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immune Cell Isolation from Mouse Femur Bone Marrow.
Liu X and Quan X.
Bio-protocol, 5(20) (2015)
Relation between phosphate energy donors and incorporation of labeled amino acids into proteins.
P C ZAMECNIK et al.
The Journal of biological chemistry, 209(1), 337-354 (1954-07-01)
Daniel L Belavý et al.
Journal of applied physiology (Bethesda, Md. : 1985), 111(5), 1304-1314 (2011-07-30)
As part of the nutrition-countermeasures (NUC) study in Cologne, Germany in 2010, seven healthy male subjects underwent 21 days of head-down tilt bed rest and returned 153 days later to undergo a second bout of 21-day bed rest. As part
Lisa Ceglia et al.
The Journal of clinical endocrinology and metabolism, 94(2), 645-653 (2008-12-04)
Protein is an essential component of muscle and bone. However, the acidic byproducts of protein metabolism may have a negative impact on the musculoskeletal system, particularly in older individuals with declining renal function. We sought to determine whether adding an
Karalee Jarvis et al.
Environmental science & technology, 43(16), 6314-6319 (2009-09-15)
Storage of CO2 through mineral sequestration using olivine has been shown to produce environmentally benign carbonates. However, due to the formation of a rate-limiting reaction product layer, the rate of reaction is insufficient for large-scale applications. We report the results

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service