Skip to Content
Merck
All Photos(1)

Key Documents

G8142

Sigma-Aldrich

Grace′s Insect Medium

With ʟ-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Grace’s Insect Media, Insect Cell Culture Medium, Insect Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

Grace′s Insect Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | insect: suitable

components

NaHCO3: 0.35 g/L
L-glutamine: 0.6 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Grace′s Insect Medium has been used:
  • in lipopolysaccharide or polysaccharide suspension preparation for administration into crickets for immune response studies
  • for resuspension of silk worm ovaries for primary cell culture preparation
  • as culture medium for bedbug for microbiome isolation

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd A Minning et al.
BMC genomics, 10, 370-370 (2009-08-12)
Chronic chagasic cardiomyopathy is a debilitating and frequently fatal outcome of human infection with the protozoan parasite, Trypanosoma cruzi. Microarray analysis of gene expression during the T. cruzi life-cycle could be a valuable means of identifying drug and vaccine targets
Oliver Otti et al.
Evolution; international journal of organic evolution, 66(3), 732-739 (2012-03-03)
Why do individuals have an imperfect immune system? Most studies suggest trade-offs associated with immunity and metabolism, and neglect ecological factors, such as predation. We provide one of the first experimental studies demonstrating a context-dependent survival cost to immune activation.
Mauricio Rv Sant'anna et al.
Parasites & vectors, 2(1), 62-62 (2009-12-17)
Leishmania parasites must overcome several barriers to achieve transmission by their sand fly vectors. One of the earliest threats is exposure to enzymes during blood meal digestion. Trypsin-like enzymes appear to be detrimental to parasite survival during the very early
CHARACTERISTICS OF VARIOUS CELL TYPES IN PRIMARY INSECT CELL CULTURE FROM LARVAL OVARIES OF SILKWORM (BOMBYX MORI L.)
Soya S, et al.
The Journal of Animal and plant sciences, 29(1), 325-332 (2019)
Regular wounding in a natural system: bacteria associated with reproductive organs of bedbugs and their quorum sensing abilities
Otti O, et al.
Frontiers in immunology, 8, 1855-1855 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service