Skip to Content
Merck
All Photos(1)

Key Documents

51942C

SAFC

Hink′s TNM-FH Insect Medium

supplemented Grace′s, with 4.1 mM L-glutamine, with 3.33 g/L lactalbumin hydrolysate (LAH), with 3.33 g/L ultrafiltered yeastolate, liquid, sterile-filtered

Synonym(s):

TNM-FH insect medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | insect: suitable

components

L-glutamine: 4.1 mM

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Hink′s TNM-FH medium was developed in 1970 for the culture of cabbagelooper, Tricoplusi in cell. Hink′s TNM-FH medium is a modified Grace′s formulation containing lactalbumin hydrolysate (LAH) and ultrafiltered yeastolate

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of media constituents on growth of insect cells in stationary and suspension cultures.
Hink, W.F., et al.
In Vitro, 9, 371-371 (1974)
Established insect cell line from the cabbage looper, Trichoplusia ni.
W F Hink
Nature, 226(5244), 466-467 (1970-05-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service