About This Item
Recommended Products
grade
for fluorescence
form
solid
solubility
methanol: soluble
fluorescence
λex 560 nm; λem 589 nm in methanol
suitability
suitable for fluorescence
SMILES string
[O-]C(=O)c1ccccc1C2=C3C=C4CCC[N+]5=C4C(CCC5)=C3Oc6c7CCCN8CCCc(cc26)c78
InChI
1S/C32H30N2O3/c35-32(36)22-10-2-1-9-21(22)27-25-17-19-7-3-13-33-15-5-11-23(28(19)33)30(25)37-31-24-12-6-16-34-14-4-8-20(29(24)34)18-26(27)31/h1-2,9-10,17-18H,3-8,11-16H2
InChI key
MUSLHCJRTRQOSP-UHFFFAOYSA-N
Related Categories
General description
Application
- as a reference model for analysing extra fluorochromic properties of rhodamine derivatives
- as a reference against thioflavin t (ThT) in the determination of ensemble photophysical properties
- as reference compound for comparing the quantum yields of nanocrystals
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service