Skip to Content
Merck
All Photos(3)

Key Documents

P9269

Millipore

Protein A–Agarose

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

extent of labeling

~2 mg per mL

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

≥10 mg/mL binding capacity (human IgG)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This Protein A-Agarose product is a conjugate of Protein A to CNBr-activated agarose without an intermediate spacer arm.

Application

Protein A-agarose is used for affinity chromatography, antibody purification and characterization, and protein A, G and L resins. Protein A-agarose has been used to study the effects of protein A immunoadsorption in patients with chronic dilated cardiomyopathy as well as to study multiple sclerosis and gastric cancer.

Quantity

Swelling: 1 g swells to approx. 4 ml

Physical form

Supplied as lyophilized powder stabilized with lactose.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Immune regulation in murine schistosomiasis japonica: inhibition of in vitro antigen- and mitogen-induced cellular responses by splenocyte culture supernatants and by purified fractions from serum of chronically infected mice.
K S Garb et al.
Journal of immunology (Baltimore, Md. : 1950), 129(6), 2752-2758 (1982-12-01)
Carl F Fortin et al.
International immunology, 22(8), 637-649 (2010-05-27)
Polymorphonuclear neutrophils (PMNs) are the first line of defense against invading organisms in humans; in addition, PMNs contribute to the linking of innate and adaptive immunity. To fulfill their biological behavior, PMNs utilize an arsenal of proteolytic enzymes, including members
Stefania Trazzi et al.
PloS one, 4(6), e5832-e5832 (2009-06-09)
CENP-C is a fundamental component of functional centromeres. The elucidation of its structure-function relationship with centromeric DNA and other kinetochore proteins is critical to the understanding of centromere assembly. CENP-C carries two regions, the central and the C-terminal domains, both
Andreas O Doesch et al.
Journal of clinical apheresis, 25(6), 315-322 (2010-09-09)
The objective of this study was to investigate functional effects of immunoadsorption (IA) in patients with chronic nonfamilial dilated cardiomyopathy (DCM) regarding clinical and humoral markers of heart failure. IA has been shown to induce early hemodynamic improvement in patients
Yu Ya Kit et al.
Biochemistry. Biokhimiia, 73(8), 950-956 (2008-09-09)
Immunoglobulins IgG and sIgA actively hydrolyzing histone H1 have been detected on analyzing proteolytic activity of antibodies isolated by chromatography on Protein A-agarose from blood serum of patients with multiple sclerosis and from colostrum of healthy mothers. These antibodies hydrolyze

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service