Skip to Content
Merck
All Photos(1)

Documents

P3391

Millipore

Protein A-Sepharose from Staphylococcus aureus

lyophilized powder

Synonym(s):

Protein A–Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

lyophilized powder

packaging

glass bottle of 1 g
glass bottle of 1.5 g
glass bottle of 250 mg
glass bottle of 5 g

storage condition

do not freeze

extent of labeling

3 mg per mL

technique(s)

affinity chromatography: suitable

color

white to gray

matrix

Sepharose CL-4B

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

≥16 mg/mL binding capacity (human IgG)

solubility

water: 1 g/L at 20 °C

suitability

conforms to structure for swelling capacity (1g swells to 4 to 5 mL)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Protein A-Sepharose when linked with antibody, helps improving antigen binding capacity of IgG crosslinked to column matrices. In this case, binding happens between Protein A and Fc portion of the IgG molecule leaving antigen specific sites free.

Application

Protein A-Sepharose was used:
  • for immunoprecipitated placentas for protein and RNA analyses using Western blot technique.
  • in chromatin immunoprecipitation to understand the link between Mediator and remodelling of chromatin structure at specific promoters.
  • as lysis buffer to remove endogenous G-type Igs in protein extraction of brain samples from female rats.
  • in immunoprecipitation of [32P] phosphomyristin C of rabbit antiserum, to study the down-regulation of protein kinase C (PKC) and phosphomyristin C (PMC) during T-cell development.
Protein A-Sepharose is used for affinity chromatography, antibody purification and characterization, immunoaffinity matrices, protein chromatography, protein A, G and L resins, and recombinant protein expression and analysis. Protein A-Sepharose has been used to develop strategies for investigating protein interactions, to improve the detection of celiac disease, and to study diabetes in children.

Quantity

Swelling: 1 g swells to approx. 4 ml.

Physical form

Supplied as lyophilized powder stabilized with lactose and dextran.

Legal Information

Sepharose is a trademark of Cytiva

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Ossipow et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(17), 8219-8223 (1993-09-01)
The CCAAT/enhancer-binding protein (C/EBP) alpha is a leucine zipper protein that is preferentially expressed in certain cell types, such as adipocytes and hepatocytes. Here we show that C/EBP alpha mRNA is translated into two major proteins, C/EBP-42 and C/EBP-30, that
Lionel Gissot et al.
Plant physiology, 142(3), 931-944 (2006-10-10)
The sucrose nonfermenting-1 protein kinase (SNF1)/AMP-activated protein kinase subfamily plays a central role in metabolic responses to nutritional and environmental stresses. In yeast (Saccharomyces cerevisiae) and mammals, the beta- and gamma-noncatalytic subunits are implicated in substrate specificity and subcellular localization
M Iezzi et al.
Molecular endocrinology (Baltimore, Md.), 13(2), 202-212 (1999-02-11)
Insulin-secreting cells express four GTPases of the Rab3 family. After separation of extracts of INS-1 cells on a sucrose density gradient, the bulk of the A, B, and C isoforms was recovered in the fractions enriched in insulin-containing secretory granules.
P Hornbeck et al.
Molecular and cellular biology, 9(9), 3727-3735 (1989-09-01)
The regulation and expression of protein kinase C (PKC) and phosphomyristin C (PMC) (a principal substrate of PKC which is the major myristylated protein in lymphocyte and glioma lines that express it) in murine B and T lymphocytes were investigated.
Tracie A Hennen-Bierwagen et al.
Plant physiology, 149(3), 1541-1559 (2009-01-27)
Starch biosynthetic enzymes from maize (Zea mays) and wheat (Triticum aestivum) amyloplasts exist in cell extracts in high molecular weight complexes; however, the nature of those assemblies remains to be defined. This study tested the interdependence of the maize enzymes

Articles

Antibody fragmentation with our pepsin digestion protocol for IgG antibody fragmentation and preparation of F(ab’).

Protocols

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service