Skip to Content
Merck
All Photos(3)

Key Documents

P7700

Millipore

Protein G–Agarose

lyophilized powder, Contains lactose stabilizers that must be removed prior to use.

Synonym(s):

Protein G resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

form

lyophilized powder

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

≥15 mg/mL binding capacity (human IgG)

storage temp.

−20°C

Application

Protein G-agarose is used in affinity chromatography, protein chromatography, antibody purification and characterization, immunoaffinity matrices, protein A, G and L resins, and purification and detection. Protein G-agarose has been used to study breast cancer and falsely elevated thyroid-stimulating hormone (TSH).

Preparation Note

Prepared with a genetically engineered Protein G which retains its high affinity for IgG and lacks albumin and Fab binding sites and membrane binding regions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Priest et al.
Molecular and biochemical parasitology, 113(1), 117-126 (2001-03-20)
Cryptosporidium parvum is a protozoan parasite of the intestinal epithelium that has caused numerous outbreaks of diarrheal illness in humans. During our studies of the host immune response to C. parvum infection, we noted that two of the immunodominant surface
John Collins et al.
Liver, 22(6), 501-506 (2002-11-26)
Persistent elevation of aspartate aminotransferase (AST) activity in serum due to the presence of a macroenzyme form of AST (macro-AST) may lead to diagnostic confusion in many clinical conditions, particularly those associated with chronic liver disease. We describe a case
Hiroshi Hirai et al.
Investigational new drugs, 29(4), 534-543 (2010-01-20)
Deregulation of cell-cycle control is a hallmark of cancer. Thus, cyclin-dependent kinases (Cdks) are an attractive target for the development of anti-cancer drugs. Here, we report the biological characterization of a highly potent pan-Cdk inhibitor with a macrocycle-quinoxalinone structure. Compound
Ebany J Martinez-Finley et al.
Neurotoxicology and teratology, 33(5), 530-537 (2011-07-26)
Changes within the glucocorticoid receptor (GR) cellular signaling pathway were evaluated in adolescent mice exposed to 50 ppb arsenic during gestation. Previously, we reported increased basal plasma corticosterone levels, decreased hippocampal GR levels and deficits in learning and memory performance
Sergueï O Fetissov et al.
Nutrition (Burbank, Los Angeles County, Calif.), 24(4), 348-359 (2008-02-12)
Peptide hormones synthesized in gastrointestinal and adipose tissues in addition to neuropeptides regulate appetite and body weight. Previously, autoantibodies directed against melanocortin peptides were found in patients with eating disorders; however, it remains unknown whether autoantibodies directed against other appetite-regulating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service