Skip to Content
Merck
All Photos(1)

Documents

P2545

Millipore

Protein A–Agarose

saline suspension

Synonym(s):

Protein A resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

biological source

Staphylococcus aureus

form

saline suspension

extent of labeling

~2 mg per mL

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

≥15 mg/mL binding capacity (human IgG)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Protein A is a 42-kDa protein from Staphylococcus aureus and has five immunoglobulin binding domains.

Application

Protein A−Agarose has been used in the immunoprecipitation
  • of protein phosphatase and tensin homolog (PTEN) protein from S63 and NIH3T3 fibroblast cells
  • of prolyl hydroxylase domain 2 N27 cells
  • of Wnt3a and sonic hedgehog (Shh) in mouse fibroblast L-cells and human embryonic kidney (HEK) 293 cells

Protein A-agarose is used for affinity chromatography, antibody purification and characterization, and protein A, G and L resins. Protein A-agarose has been used to study the effects of protein A immunoadsorption in patients with chronic dilated cardiomyopathy as well as to study multiple sclerosis and gastric cancer.

Biochem/physiol Actions

Protein A binds to Fcγ (the constant region) and Fab (antigen binding fragment) of IgG. Protein A agarose resin is commercially used for affinity based purifications.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTEN-Dependent Stabilization of MTSS1 Inhibits Metastatic Phenotype in Pancreatic Ductal Adenocarcinoma
Zeleniak AE, et al.
Neoplasia, 20(1), 12-24 (2018)
D J McCool et al.
The Biochemical journal, 302 ( Pt 1), 111-118 (1994-08-15)
Pulse-chase labelling experiments were performed using the mucin-producing colonic carcinoma cell line LS180. Cells were pulsed with [3H]threonine or [3H]glucosamine and chased in complete media without radiolabel for various lengths of time. From cell and media extracts obtained at each
Membrane targeting of palmitoylated Wnt and Hedgehog revealed by chemical probes
Gao X, et al.
Febs Letters, 585(15), 2501-2506 (2011)
Madhu Chhanda Mohanty et al.
Developmental and comparative immunology, 26(4), 385-391 (2002-03-13)
Meriones unguiculatus commonly known as gerbils are widely used as animal models for a variety of parasitic infections such as Brugia malayi, Entamoeba histolytica, Giardia duodenalis, Toxoplasma gondi, Helicobacter pylori, Strongyloides stercoralis and Echinococcus multilocularis. Groups of BALB/c mice, gerbils
Hsp90 Co-chaperone p23 contributes to dopaminergic mitochondrial stress via stabilization of PHD2: Implications for Parkinson?s disease
Rane A, et al.
Neurotoxicology, 65, 166-173 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service