Skip to Content
Merck
All Photos(3)

Key Documents

P9053

Sigma-Aldrich

Pyridine-2-aldoxime methochloride

Synonym(s):

2-PAM chloride, Pralidoxime chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N2O · Cl
CAS Number:
Molecular Weight:
172.61
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

form

solid

mp

230 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

SMILES string

[Cl-].C[n+]1ccccc1\C=N\O

InChI

1S/C7H8N2O.ClH/c1-9-5-3-2-4-7(9)6-8-10;/h2-6H,1H3;1H

InChI key

HIGSLXSBYYMVKI-UHFFFAOYSA-N

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

The prototypical reactivator of acetylcholinesterase that has been inactivated by organophosphorus insecticides or nerve agents. It is now known that no reactivator is effective against a broad spectrum of organophosphorus agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamil Musilek et al.
Bioorganic & medicinal chemistry, 16(17), 8218-8223 (2008-08-05)
Acetylcholinesterase reactivators are crucial antidotes for the treatment of organophosphate intoxication. Eighteen monoquaternary reactivators of acetylcholinesterase with modified side chain were developed in an effort to extend the properties of pralidoxime. The known reactivators (pralidoxime, HI-6, obidoxime, trimedoxime, methoxime) and
Jyotiranjan Acharya et al.
European journal of medicinal chemistry, 46(9), 3926-3933 (2011-06-28)
A series of carbamoyl bis-pyridinium monooximes linked with xylene linker were synthesized and their in-vitro reactivation potential was evaluated against acetylcholinesterase (AChE) inhibited by organophosphorus inhibitors (OP) such as sarin, DFP and VX and the data were compared with reactivation
Chennamaneni Srinivas Rao et al.
Bioorganic & medicinal chemistry letters, 16(8), 2134-2138 (2006-02-17)
In continuation of our investigations of unsymmetrical bisquaternary monooximes, we synthesized four new series of compounds bridged by hexyl, heptyl, octyl and nonyl groups. All eight monooximes viz., dibromides of 1-(4-hydroxyiminomethylpyridinium)6-(3/4-carbamoylpyridinium)hexane, 1-(4-hydroxyiminomethylpyridinium)-7-(3/4-carbamoylpyridinium)heptane, 1-(4-hydroxyiminomethylpyridinium)-8-(3/4-carbamoylpyridinium)octane, 1-(4-hydroxyiminomethylpyridinium)-9-(3/4-carbamoylpyridinium)nonane as well as the corresponding bis-oximes
Kamil Musilek et al.
Bioorganic & medicinal chemistry letters, 16(21), 5673-5676 (2006-08-29)
Three asymmetrical AChE reactivators with cyano-moiety and propane linker were synthesized using modification of currently known synthetic pathways. Their potency to reactivate AChE inhibited by nerve agent tabun and insecticide paraoxon was tested in vitro and compared to pralidoxime, HI-6
Kamil Musilek et al.
Bioorganic & medicinal chemistry letters, 17(11), 3172-3176 (2007-03-27)
Six novel AChE reactivators with a (Z)-but-2-ene linker were synthesized using the known synthetic pathways. Their ability to reactivate AChE, which had been previously inhibited by nerve agent tabun or pesticide paraoxon, was tested in vitro and compared to pralidoxime

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service