Skip to Content
Merck
All Photos(3)

Key Documents

P62003

Sigma-Aldrich

2-Pyridinecarboxaldehyde

99%

Synonym(s):

Picolinaldehyde, Pyridine-2-aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO
CAS Number:
Molecular Weight:
107.11
Beilstein:
105341
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%
Pricing and availability is not currently available.

Quality Level

Assay

99%

refractive index

n20/D 1.536 (lit.)

bp

181 °C (lit.)

density

1.126 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccccn1

InChI

1S/C6H5NO/c8-5-6-3-1-2-4-7-6/h1-5H

InChI key

CSDSSGBPEUDDEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Pyridinecarboxaldehyde has been used:
  • To synthesize chitosan based bifunctionalized adsorbent.[1]
  • In the aldol addition reaction with pyruvate in the presence of 2-keto-3-deoxy-6-phosphogluconate aldolase (KDPG) catalyst to form (S)-4-hydroxy-2-keto-4-(2′-pyridyl)butyrate.[2]
  • In the condensation reaction with (S)-(−)-α-methylbenzylamine and (R)-(+)-α-methylbenzylamine to synthesize two chiral (S)-(−)- and (R)-(+)Schiff base compounds.[3]

It can also be combined with thiosemicarbazide to form 2-pyridinecarboxaldehyde thiosemicarbazone ligand (L) which can further complex with Ni(II) and Cu(II) salts (MX) to form [M(L)2X2] complexes.[4]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

170.6 °F

Flash Point(C)

77 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bifunctionalized chitosan: A versatile adsorbent for removal of Cu (II) and Cr (VI) from aqueous solution
Moreira ALSL, et al.
Carbohydrate Polymers, 201, 218-227 (2018)
Bacterial biodegradability of inhibitors of chiral imine type corrosion and catabolic prediction
Bautista Garcia JE, et al.
REVISTA INTERNACIONAL DE CONTAMINACION AMBIENTAL, 31(4), 415-426 (2015)
Olga Yu Vassilyeva et al.
Dalton transactions (Cambridge, England : 2003), 48(30), 11278-11284 (2019-07-04)
The novel organic-inorganic hybrid salt [L]2[CoCl4] (1) and molecular complex [CoLCl3] (2), where L+ is 2-methyl-3-(pyridin-2-yl)imidazo[1,5-a]pyridinium cation, feature simple {CoCl4} and {CoCl3N} tetrahedral environments of negligible (1) and a slightly higher distortion (2) that are responsible for rather low positive
Ferenc Matyuska et al.
Dalton transactions (Cambridge, England : 2003), 46(26), 8626-8642 (2017-06-27)
Manganese(ii), copper(ii) and zinc(ii) complexes of four polydentate tripodal ligands (tachpyr (N,N',N''-tris(2-pyridylmethyl)-cis,cis-1,3,5-triaminocyclohexane), trenpyr (tris[2-(2-pyridylmethyl)aminoethyl]amine, tach3pyr (N,N',N''-tris(3-pyridylmethyl)-cis,cis-1,3,5-triaminocyclohexane) and tren3pyr (tris[2-(2-pyridylmethyl)aminoethyl]amine)) were characterized in both solution and solid states. A combined evaluation of potentiometric, UV-VIS, NMR and EPR data allowed the conclusion
Gaganpreet Kaur et al.
Dalton transactions (Cambridge, England : 2003), 47(16), 5595-5606 (2018-03-17)
The unregulated use of organophosphates (OPs) as pesticides and toxic chemical warfare agents demands their continuous monitoring from a human health perspective. This study describes a fluorescence turn-on sensing assay for the selective quantification of OPs in aqueous medium. Metal

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service