Skip to Content
Merck
All Photos(1)

Documents

A0257

Sigma-Aldrich

Atropine sulfate salt monohydrate

≥97% (TLC), crystalline, cholinergic receptor antagonist

Synonym(s):

α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, Tropine tropate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H23NO3)2 · H2SO4 · H2O
CAS Number:
Molecular Weight:
694.83
Beilstein:
6109275
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.32

product name

Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline

Assay

≥97% (TLC)

form

crystalline

color

white

mp

189-192 °C (A) (lit.)

solubility

H2O: 2.5 g/mL (stable for several days at 4 °C.)
ethanol: 200 mg/mL (stable for several days at 4 °C.)
glycerol: 400 mg/mL (stable for several days at 4 °C.)

SMILES string

O.OS(O)(=O)=O.CN1[C@H]2CC[C@@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3.CN4[C@H]5CC[C@@H]4C[C@@H](C5)OC(=O)C(CO)c6ccccc6

InChI

1S/2C17H23NO3.H2O4S.H2O/c2*1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12;1-5(2,3)4;/h2*2-6,13-16,19H,7-11H2,1H3;(H2,1,2,3,4);1H2/t2*13-,14+,15+,16?;;

InChI key

JPKKQJKQTPNWTR-CHYDPLAESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atropine is isolated from Atropa belladonna, Datura stramonium and Mandragora officinarum. It is a tertiary amine, which acts as a cholinergic receptor antagonist.

Application

Atropine sulfate salt hydrate has been used:
  • to assay specific binding of high-affinity 3-quinuclidinyl benzilate-3H ([3H]QNB) to the muscarinic acetylcholine receptor (mAChR)
  • to block the recovery phase of the aldicarb (Ald) response
  • as an analyte for liquid chromatography with tandem mass spectrometry (LC-MS/MS)
  • as a mAChR antagonist to block acetylcholine receptor

Biochem/physiol Actions

Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Acute effects of drugs on C. elegans movement reveal complex responses and plasticity
Spensley MA, et al.
G3 (Bethesda, Md.), 299719-299719 (2018)
Method validation for simultaneous determination of atropine, pralidoxime and 12 organophosphorus compounds in blood samples by means of high-performance liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
<BIG>Marques GLM, et al</BIG>.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1097, 44-53 (2018)
Spinalization and locomotor training differentially affect muscarinic acetylcholine receptor type 2 abutting on alpha-motoneurons innervating the ankle extensor and flexor muscles
Wikeckowska A, et al.
Journal of Neurochemistry, 147(3), 361-379 (2018)
Faust's Anesthesiology Review E-Book (2014)
Laxmikant S Deshpande et al.
Toxicological sciences : an official journal of the Society of Toxicology, 116(2), 623-631 (2010-05-26)
Organophosphate (OP) compounds are among the most lethal chemical weapons ever developed and are irreversible inhibitors of acetylcholinesterase. Exposure to majority of OP produces status epilepticus (SE) and severe cholinergic symptoms that if left untreated are fatal. Survivors of OP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service