Skip to Content
Merck
All Photos(1)

Key Documents

P5296

Sigma-Aldrich

CTOP

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H67N11O11S2
CAS Number:
Molecular Weight:
1062.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

C[C@@H](O)[C@H](NC(=O)[C@H]1NC(=O)[C@@H](NC(=O)[C@H](CCCN)NC(=O)[C@@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@H](CSSC1(C)C)NC(=O)[C@H](N)Cc5ccccc5)[C@@H](C)O)C(N)=O

InChI

1S/C50H67N11O11S2/c1-26(62)39(42(53)65)59-49(72)41-50(3,4)74-73-25-38(58-43(66)33(52)21-28-11-6-5-7-12-28)47(70)56-36(22-29-16-18-31(64)19-17-29)45(68)57-37(23-30-24-54-34-14-9-8-13-32(30)34)46(69)55-35(15-10-20-51)44(67)60-40(27(2)63)48(71)61-41/h5-9,11-14,16-19,24,26-27,33,35-41,54,62-64H,10,15,20-23,25,51-52H2,1-4H3,(H2,53,65)(H,55,69)(H,56,70)(H,57,68)(H,58,66)(H,59,72)(H,60,67)(H,61,71)/t26-,27-,33-,35+,36+,37-,38+,39+,40+,41-/m1/s1

InChI key

PZWWYAHWHHNCHO-FGHAYEPSSA-N

Gene Information

mouse ... Pnoc(18155)

Amino Acid Sequence

Phe-Cys-Tyr-Trp-Orn-Thr-Pen-Thr-NH2 [Disulfide Bridge: 2-7]

General description

D-Pen-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH2 (CTOP) is a selective cyclic μ-opioid receptor antagonist.

Application

CTOP has been used:
  • to study the anxiogenic effects induced by CTOP in mice and rat.
  • to study the effect of μ-opioid antagonist, CTOP on the bovine milk-derived LF (BLF)-induced analgesia.
  • to determine whether μ-opioid receptors act cooperatively with 5-hydroxytryptamine (5-HT1A) receptors to regulate the behaviors generated in the elevated T-maze (ETM).

Biochem/physiol Actions

D-Pen-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH2 (CTOP) inhibits acute morphine-induced hypermotility and analgesic effect of morphine in a dose-dependent manner in vivo.
Selective ligand for μ-opioid receptors.

Linkage

Somatostatin analog

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena I Varlinskaya et al.
Alcoholism, clinical and experimental research, 33(6), 991-1000 (2009-03-24)
Ethanol consumption is considerably elevated during adolescence. Attractiveness of alcohol for humans during the adolescent developmental period is based, in part, on its ability to induce social facilitation--a facilitation of social interactions not only evident in human adolescents but also
Ruth Drdla-Schutting et al.
Science (New York, N.Y.), 335(6065), 235-238 (2012-01-17)
Painful stimuli activate nociceptive C fibers and induce synaptic long-term potentiation (LTP) at their spinal terminals. LTP at C-fiber synapses represents a cellular model for pain amplification (hyperalgesia) and for a memory trace of pain. μ-Opioid receptor agonists exert a
Chronic pain induces anxiety with concomitant changes in opioidergic function in the amygdala.
Narita M
Neuropsychopharmacology, 31(4), 739-750 (2006)
Lactoferrin enhances opioid-mediated analgesia via nitric oxide in the rat spinal cord.
Hayashida K
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology, 285(2), R306-R312 (2003)
Kate M Wassum et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(5), 1591-1599 (2011-02-04)
The decision to perform, or not perform, actions known to lead to a rewarding outcome is strongly influenced by the current incentive value of the reward. Incentive value is largely determined by the affective experience derived during previous consumption of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service