Skip to Content
Merck
All Photos(1)

Documents

C2901

Sigma-Aldrich

Cholecystokinin (CCK) Fragment 26-33 Amide, Non-sulfated

≥95% (HPLC), powder

Synonym(s):

CCK-8, non-sulfated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H62N10O13S2
CAS Number:
Molecular Weight:
1063.21
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(N)CC(O)=O)C(=O)NCC(=O)NC(Cc2c[nH]c3ccccc23)C(=O)NC(CCSC)C(=O)NC(CC(O)=O)C(=O)NC(Cc4ccccc4)C(N)=O

InChI

1S/C49H62N10O13S2/c1-73-18-16-34(55-47(70)37(21-28-12-14-30(60)15-13-28)58-44(67)32(50)23-41(62)63)45(68)53-26-40(61)54-38(22-29-25-52-33-11-7-6-10-31(29)33)48(71)56-35(17-19-74-2)46(69)59-39(24-42(64)65)49(72)57-36(43(51)66)20-27-8-4-3-5-9-27/h3-15,25,32,34-39,52,60H,16-24,26,50H2,1-2H3,(H2,51,66)(H,53,68)(H,54,61)(H,55,70)(H,56,71)(H,57,72)(H,58,67)(H,59,69)(H,62,63)(H,64,65)

InChI key

OIXQINQYMGNCII-UHFFFAOYSA-N

Gene Information

human ... CCK(885)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Asp-Tyr-Met-Gly-Trp-Met-Asp-Phe-NH2

Application

Cholecystokinin (CCK) Fragment 26-33 Amide, Non-sulfated has been used in the specificity testing against gastrin and pepsinogen I proteins.

Biochem/physiol Actions

Biologically inactive form of CCK-8; CCKB/gastrin receptor agonist that has no activity at the CCKA receptor.
Cholecystokinin (CCK) is a 115 amino acid preprohormone and a neurotransmitter. It exists in many isoforms. The octapeptide of CCK (CCK-8S) is the most abundant form. CCK is mapped to human chromosome 3p22.1. CCK interacts with receptors of the gastrointestinal tract and nervous system. Polymorphisms in the CCK is implicated in hallucinations in patients with Parkinson′s disease. CCK plays a key role in the activation of the calcineurin-regulated nuclear factor of activated T-cells (NFATs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Genetic substrates of psychosis in patients with Parkinson's disease: a critical review.
Lenka A, et al.
Journal of the Neurological Sciences, 364(5), 33-41 (2016)
Atrophic gastritis serum levels of amidated gastrin-17 and pepsinogen I in atrophic gastritis: an observational case-control study.
Sipponen P, et al.
Scandinavian Journal of Gastroenterology, 37(7), 785-791 (2002)
Advances in molecular genetics of panic disorder.
Maron E, et al.
Molecular Psychiatry, 15(7), 681-681 (2010)
Neuropeptide regulation of fear and anxiety: Implications of cholecystokinin, endogenous opioids, and neuropeptide γ.
Bowers ME, et al.
Physiology & Behavior, 107(5), 699-710 (2012)
A Maeda et al.
Pharmacological research, 42(4), 333-336 (2000-09-16)
The sulfated and unsulfated forms of cholecystokinin-octapeptide (CCK-8) were compared, with respect to their effect on gastric acid secretion, in the rat. Unsulfated CCK-8 stimulated acid secretion in a dose-dependent manner, while the sulfated form was without stimulatory effect; thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service