Skip to Content
Merck
All Photos(3)

Documents

595063

Sigma-Aldrich

Titanium nitride

<3 μm

Synonym(s):

Titanium mononitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiN
CAS Number:
Molecular Weight:
61.87
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

powder

particle size

<3 μm

mp

2930 °C (lit.)

density

5.24 g/mL at 25 °C (lit.)

SMILES string

N#[Ti]

InChI

1S/N.Ti

InChI key

NRTOMJZYCJJWKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chuan Ting Wang et al.
Journal of the mechanical behavior of biomedical materials, 17, 166-175 (2012-11-13)
A commercial purity (CP) Grade 2 Ti was processed by high-pressure torsion (HPT) using an imposed pressure of 3.0GPa at room temperature. The HPT processing reduced the grain size from ∼8.6 μm in the as-received state to ultra-fine grains (UFG)
P R Mezger et al.
Journal of dentistry, 20(6), 342-344 (1992-12-01)
Titanium and its alloys are increasingly important in dentistry. Thin titanium nitride (TiN) coatings improve properties of metallic material for industrial purposes. Recently TiN coatings have been advocated in dentistry. In a survey of the literature, aspects of biological, mechanical
Duck Hyun Youn et al.
ChemSusChem, 6(2), 261-267 (2013-01-11)
The development of an efficient noble-metal-free counter electrode is crucial for possible applications of quantum-dot-sensitized solar cells (QDSSCs). Herein, we present TiN nanoparticles on a carbon nanotube (CNT)-graphene hybrid support as a noble-metal-free counter electrode for QDSSCs employing a polysulfide
Aleksandra Dubiel et al.
Materials (Basel, Switzerland), 13(5) (2020-03-04)
In this work, two kinds of silicon nitride-based composites, namely, those with titanium nitride or silicon carbide additives, were sintered using the hot pressing technique (HP). The phase composition, microstructure, and mechanical and elastic properties of the materials were characterized.
Michele C B de Castro et al.
Materials (Basel, Switzerland), 12(3) (2019-02-13)
The Ti-6Al-4V alloy is widely used in the manufacture of components that must have low density and high corrosion resistance and fatigue strength. The fatigue strength can be improved by surface modification. The aim of this study was to determine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service