Skip to Content
Merck
All Photos(1)

Key Documents

C6352

Sigma-Aldrich

CTAP

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H69N13O11S2
CAS Number:
Molecular Weight:
1104.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95%

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

C[C@@H](O)[C@H](NC(=O)[C@H]1NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@H](CSSC1(C)C)NC(=O)[C@H](N)Cc5ccccc5)[C@@H](C)O)C(N)=O

InChI

1S/C51H69N13O11S2/c1-26(65)39(42(53)68)62-49(75)41-51(3,4)77-76-25-38(61-43(69)33(52)21-28-11-6-5-7-12-28)47(73)59-36(22-29-16-18-31(67)19-17-29)45(71)60-37(23-30-24-57-34-14-9-8-13-32(30)34)46(72)58-35(15-10-20-56-50(54)55)44(70)63-40(27(2)66)48(74)64-41/h5-9,11-14,16-19,24,26-27,33,35-41,57,65-67H,10,15,20-23,25,52H2,1-4H3,(H2,53,68)(H,58,72)(H,59,73)(H,60,71)(H,61,69)(H,62,75)(H,63,70)(H,64,74)(H4,54,55,56)/t26-,27-,33-,35+,36+,37-,38+,39+,40+,41-/m1/s1

InChI key

OFMQLVRLOGHAJI-FGHAYEPSSA-N

Gene Information

rat ... Pnoc(25516)

Amino Acid Sequence

D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH2

Application

CTAP has been used as a μ-opioid receptor (MOR) antagonist:
  • to study the anti-hyperalgesic effect of dipeptidyl peptidase 4 (DPP4) inhibitor isoleucine-proline-isoleucine (IPI) and vildagliptin in carrageenan-induced inflammation
  • to study the role of MOR in glutamate and gamma-aminobutyric acid (GABA) efflux during predator stress in rats
  • to determine the endogenous opioid peptide involved in blocking pain induced by activated gastrin-releasing peptide (Grp+) neurons

Biochem/physiol Actions

CTAP is a peptide antagonist produced from somatostatin analogs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Abbruscato et al.
The Journal of pharmacology and experimental therapeutics, 280(1), 402-409 (1997-01-01)
D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH2 (CTAP) is a cyclic, penicillamine-containing octapeptide that is structurally similar to somatostatin and displays greater antagonist potency and selectivity for mu-opioid receptors, compared with the classical mu-selective antagonist D-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH2. The aim of this study was to determine whether CTAP
Chloe M Stanton et al.
Scientific reports, 7(1), 12147-12147 (2017-09-25)
Late-onset retinal degeneration (L-ORD) is a rare autosomal dominant retinal dystrophy, characterised by extensive sub-retinal pigment epithelium (RPE) deposits, RPE atrophy, choroidal neovascularisation and photoreceptor cell death associated with severe visual loss. L-ORD shows striking phenotypic similarities to age-related macular
Leaky gate model: intensity-dependent coding of pain and itch in the spinal cord
Sun S, et al.
Neuron, 93(4), 840-853 (2017)
Kevin Caref et al.
eLife, 7 (2018-03-28)
When relatively sated, people (and rodents) are still easily tempted to consume calorie-dense foods, particularly those containing fat and sugar. Consumption of such foods while calorically replete likely contributes to obesity. The nucleus accumbens (NAc) opioid system has long been
E J Bilsky et al.
The Journal of pharmacology and experimental therapeutics, 277(1), 484-490 (1996-04-01)
Previous studies measuring opioid inhibition of cyclic adenosine monophosphate in SH-SY5Y cells supported the hypothesis that continuous agonist stimulation causes a gradual conversion of the mu opioid receptor to a sensitized or constitutively active state termed mu*. Conversion to mu*

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service