Skip to Content
Merck
All Photos(1)

Key Documents

P5368

Sigma-Aldrich

Phosphate buffered saline

powder, pH 7.4, BioPerformance Certified, for molecular biology

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
NACRES:
NA.25
Pricing and availability is not currently available.

Product Name

Phosphate buffered saline, BioPerformance Certified, pH 7.4

grade

BioPerformance Certified
for molecular biology

Quality Level

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

technique(s)

immunofluorescence: suitable

impurities

BioBurden, tested
endotoxin, tested

color

white

pH

7.4

solubility

water: 9.07-10.0 g/L, clear, colorless

cation traces

Fe: ≤10 ppm
heavy metals (as Pb): ≤5 ppm

suitability

suitable for SDS-PAGE
suitable for molecular biology

application(s)

diagnostic assay manufacturing
histology
sample preparation

foreign activity

DNase, RNase, Protease, none detected

SMILES string

OP(=O)(O)[O-].OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Cl-].[Cl-].[K+].[K+]

InChI

InChI=1S/2ClH.2K.3Na.2H3O4P/c;;;;;;;2*1-5(2,3)4/h2*1H;;;;;;2*(H3,1,2,3,4)/q;;5*+1;;/p-5

InChI key

LOKCTEFSRHRXRJ-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application


  • Rapid and zero-cost DNA extraction from soft-bodied insects for routine PCR-based applications.: This study used phosphate buffered saline (PBS) to develop a cost-effective and efficient method for DNA extraction from insects, facilitating routine PCR-based applications in molecular biology (Jangra & Ghosh, 2022).

  • Injectable amnion hydrogel-mediated delivery of adipose-derived stem cells for osteoarthritis treatment.: PBS was utilized to prepare and deliver adipose-derived stem cells in an injectable hydrogel, showing potential for innovative osteoarthritis treatments (Bhattacharjee et al., 2022).

  • Rapid and sensitive detection of SARS-CoV-2 infection using quantitative peptide enrichment LC-MS analysis.: This research employed PBS for sample preparation in a novel LC-MS method for detecting SARS-CoV-2, providing a rapid and sensitive diagnostic tool (Hober et al., 2021).

Packaging

Foil Pouch

Reconstitution

Contents of one pouch, when dissolved in one liter of distilled or deionized water, will yield 0.01 M phosphate buffered saline (NaCl 0.138 M; KCl - 0.0027 M); pH 7.4, at 25 °C.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petra J Roubos-van den Hil et al.
Veterinary microbiology, 202, 58-63 (2016-09-27)
Enterotoxigenic E. coli (ETEC), causing post-weaning diarrhoea, is a major problem in weaned piglets. Individual animal responses to ETEC infection show high variability in animal experiments. Two studies were designed to optimize the ETEC F4ac infection model in piglets by
Jesús Gilberto Arámburo-Galvez et al.
Nutrients, 10(7) (2018-07-18)
Most food allergy cases are associated with a limited group of allergens. This could be attributed to an increased ability of some foods to sensitize and trigger allergic reactions. However, there are no validated animal models to evaluate the sensitizing
Jinjin Sun et al.
Nanoscale research letters, 14(1), 248-248 (2019-07-26)
Herein, a magnetic graphene field-effect transistor biosensor was prepared through the transfer of a chemical vapor deposition graphene film onto a glass substrate to produce a sensing film and conductive channel. By fixing 1-pyrenebutanoic acid succinimidyl ester onto graphene film
Gary Burness et al.
Biology open, 7(4) (2018-04-06)
Female birds transfer maternally derived antibodies (matAb) to their nestlings, via the egg yolk. These antibodies are thought to provide passive protection, and allow nestlings to avoid the costs associated with mounting an innate immune response. To test whether there
Yixi Chen et al.
The European journal of neuroscience, 49(4), 510-524 (2018-11-26)
An emerging treatment for Parkinson's disease (PD) is cell replacement therapy. Authentic midbrain dopaminergic (mDA) neuronal precursors can be differentiated from human embryonic stem cells (hESCs) and human induced pluripotent stem cells (iPSCs). These laboratory-generated mDA cells have been demonstrated

Articles

Learn about Supel™ BioSPME C18 96-pin tool to prepare a clean sample with minimal matrix effects for an LC-MS/MS analysis from human plasma and its comparison with acetonitrile-based precipitation.

Learn about Supel™ BioSPME C18 96-pin tool to prepare a clean sample with minimal matrix effects for an LC-MS/MS analysis from human plasma and its comparison with acetonitrile-based precipitation.

Learn about Supel™ BioSPME C18 96-pin tool to prepare a clean sample with minimal matrix effects for an LC-MS/MS analysis from human plasma and its comparison with acetonitrile-based precipitation.

Learn about Supel™ BioSPME C18 96-pin tool to prepare a clean sample with minimal matrix effects for an LC-MS/MS analysis from human plasma and its comparison with acetonitrile-based precipitation.

See All

Protocols

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Questions

1–7 of 7 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the shelf-life of product P5368, phosphate buffered saline (PBS)?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

  4. What volume of Product P5368, Phosphate buffered saline (PBS), does each pack make?

    1 answer
    1. When the contents of a pouch is dissolved in 1 Liter of water, it makes 1X PBS, pH 7.4 at 25°C.

      Helpful?

  5. If reconstituted with sterile water, will Product P5368, Phosphate buffered saline (PBS), be sterile?

    1 answer
    1. No.  To make this PBS sterile, the solution should be either sterile-filtered or autoclaved.

      Helpful?

  6. Does Product P5368, Phosphate buffered saline (PBS), contain calcium and magnesium?

    1 answer
    1. No.  The complete formulation is shown in the product listing.  When reconstituted, the resulting solution is 10 mM phosphate buffer, with 138 mM sodium chloride and 2.7 mM potassium chloride.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service