Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0618-01

Protein G Sepharose 4 Fast Flow

Cytiva 17-0618-01, pack of 5 mL

Synonym(s):

Fast Flow resin, Antibody purification resin, IgG purification resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

recombinant protein G lacking albumin-binding region

packaging

pack of 5 mL

manufacturer/tradename

Cytiva 17-0618-01

storage condition

(20% Ehtanol)

matrix

4% cross-linked agarose

average diameter

90 μm (d50v)

cleaning in place

2-10

working range

3-9

suitability

suitable for bioprocess medium

storage temp.

2-8°C

General description

Protein G Sepharose 4 Fast Flow is recombinant protein G coupled to Sepharose 4 Fast Flow.

Protein G Sepharose 4 Fast Flow has recombinant protein G immobilized by the cyanogen bromide (CNBr) method to Sepharose 4 Fast Flow. Protein G exhibit binding specificities that complement Protein A media and binds to the Fc region of IgG from a variety of mammalian species. Protein G Sepharose 4 Fast Flow may be used to isolate and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.

As member of the BioProcess media range, Protein G Sepharose 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • Binding specificities that complement Protein A media.
  • Binds a broad range of IgG species and subclasses.
  • Multi-point attachment minimizes ligand leakage.
  • Used in a range of research applications.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marti Quevedo et al.
Nature communications, 10(1), 2669-2669 (2019-06-19)
The Mediator complex regulates transcription by connecting enhancers to promoters. High Mediator binding density defines super enhancers, which regulate cell-identity genes and oncogenes. Protein interactions of Mediator may explain its role in these processes but have not been identified comprehensively.
Bodan Hu et al.
Bio-protocol, 10(4), e3523-e3523 (2021-03-04)
Non-covalent binding of cholesterol to the transmembrane region of proteins affect their functionalities, but methods to prove such an interaction are rare. We describe our protocol to label the hemagglutinin (HA) of Influenza virus with a cholesterol derivative in living
Ting-Ting Du et al.
Nature communications, 10(1), 1117-1117 (2019-03-10)
Sensory hair cells, the mechanoreceptors of the auditory and vestibular systems, harbor two specialized elaborations of the apical surface, the hair bundle and the cuticular plate. In contrast to the extensively studied mechanosensory hair bundle, the cuticular plate is not
Yunhao Tan et al.
Methods in molecular biology (Clifton, N.J.), 1714, 79-95 (2017-11-28)
Ligand-induced macromolecular protein complex formation has emerged as a common means by which the innate immune system activates signal transduction pathways essential for host defense. Despite their structural divergence, key signaling molecules in diverse innate immune pathways mediate signal transduction
Hao Zheng et al.
Redox biology, 48, 102175-102175 (2021-11-05)
Ferroptosis is a form of regulated cell necrosis, as a consequence of Fe(II)-dependent lipid peroxidation. Although ferroptosis has been linked to cancer cell death, neurodegeneration and reperfusion injury, physiological roles of ferroptosis have not been elucidated to date mostly due

Articles

This page shows a comparison of the relative binding strengths of protein G and protein A to different immunoglobulins.

Purify monoclonal or polyclonal IgG from serum, cell culture supernatant or ascitic fluid using the HiTrap Protein G HP from Cytiva, an affinity-exclusion chromatography product containing Sepharose-immobilized Protein G.

This page describes immunoprecipitation (immunoaffinity or pull-down techniques).

This page describes efficient column packing and preparation for affinity chromatography of antibodies.

Protocols

This page provides information about different pull-down assays for the further isolation of multiprotein complexes to identify their components with products from Cytiva.

This page shows how to separate IgG antibodies by affinity chromatography using Protein G Sepharose 4 Fast Flow from Cytiva.

Related Content

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service