Skip to Content
Merck
All Photos(1)

Key Documents

N0630

Sigma-Aldrich

Niflumic acid

Synonym(s):

2-(α,α,α-Trifluoro-m-toluidino)nicotinic acid, 2-[3-(Trifluoromethyl)anilino]nicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9F3N2O2
CAS Number:
Molecular Weight:
282.22
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98% (HPLC)

form

powder

mp

203-204 °C

solubility

acetone: 50 mg/mL, clear to slightly hazy, yellow

SMILES string

O=C(O)C1=CC=CN=C1NC2=CC(C(F)(F)F)=CC=C2

InChI

1S/C13H9F3N2O2/c14-13(15,16)8-3-1-4-9(7-8)18-11-10(12(19)20)5-2-6-17-11/h1-7H,(H,17,18)(H,19,20)

InChI key

JZFPYUNJRRFVQU-UHFFFAOYSA-N

Gene Information

human ... PTGS1(5742)

Looking for similar products? Visit Product Comparison Guide

Application

Niflumic acid has been used:
  • as a chloride channel blocker in mouse olfactory epithelium for odor-induced electroolfactograms (EOGs) measurements, to test its analgesic effect on rat dorsal root ganglion neurons post γ-aminobutyric acid induction
  • as a calcium-activated chloride channel (CaCC) inhibitor in C2C12 myoblasts and also to test its effect on volume-regulated anion channel (VRAC)
  • as an inhibitor of voltage-dependent potassium channel in human embryonic kidney (HEK 293) phoenix cells

Biochem/physiol Actions

Niflumic acid (NFA), a γ-aminobutyric acid type A receptor (GABAARs) antagonist is a non-steroidal anti-inflammatory drug (NSAID) and it belongs to the fenamate class. It is also a blocker of chloride ion channel and a calcium-activated chloride channel (CaCC) inhibitor. NFA possesses anti-inflammatory property and is useful in treating rheumatic disorders. It is also an inhibitor of N-methyl-D-aspartate receptor and glycine receptor. NFA also inhibits enzymes associated with the prostaglandins synthesis.
Selective cyclooxygenase-2 (COX-2) inhibitor.

Features and Benefits

This compound is featured on the Chloride Channels and Potassium Channels pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Dibattista et al.
The Journal of general physiology, 140(1), 3-15 (2012-06-27)
The rodent vomeronasal organ plays a crucial role in several social behaviors. Detection of pheromones or other emitted signaling molecules occurs in the dendritic microvilli of vomeronasal sensory neurons, where the binding of molecules to vomeronasal receptors leads to the
Juraj Rievaj et al.
Pflugers Archiv : European journal of physiology, 463(3), 497-509 (2011-12-16)
Protease-activated receptor 2 (PAR-2) is a G protein-coupled receptor possibly involved in the pathogenesis of asthma. PAR-2 also modulates ion transport in cultured epithelial cells, but these effects in native airways are controversial. The influence of allergic inflammation on PAR-2-induced
Luis Alvarez et al.
The Journal of cell biology, 196(5), 653-663 (2012-03-01)
During chemotaxis and phototaxis, sperm, algae, marine zooplankton, and other microswimmers move on helical paths or drifting circles by rhythmically bending cell protrusions called motile cilia or flagella. Sperm of marine invertebrates navigate in a chemoattractant gradient by adjusting the
Franziska Neureither et al.
Physiological reports, 5(15) (2017-08-09)
Many animals follow odor trails to find food, nesting sites, or mates, and they require only faint olfactory cues to do so. The performance of a tracking dog, for instance, poses the question on how the animal is able to
Galyna Maleeva et al.
Frontiers in molecular neuroscience, 10, 125-125 (2017-06-01)
Niflumic acid (NFA) is a member of the fenamate class of nonsteroidal anti-inflammatory drugs. This compound and its derivatives are used worldwide clinically for the relief of chronic and acute pain. NFA is also a commonly used blocker of voltage-gated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service