Skip to Content
Merck
All Photos(1)

Documents

Y0000824

Niflumic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(α,α,α-Trifluoro-m-toluidino)nicotinic acid, 2-[3-(Trifluoromethyl)anilino]nicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9F3N2O2
CAS Number:
Molecular Weight:
282.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

niflumic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(O)C1=CC=CN=C1NC2=CC(C(F)(F)F)=CC=C2

InChI

1S/C13H9F3N2O2/c14-13(15,16)8-3-1-4-9(7-8)18-11-10(12(19)20)5-2-6-17-11/h1-7H,(H,17,18)(H,19,20)

InChI key

JZFPYUNJRRFVQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Niflumic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Selective cyclooxygenase-2 (COX-2) inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Cousin et al.
The Journal of membrane biology, 46(2), 125-153 (1979-04-20)
In human erythrocyte, permeability to the anion is instantaneously, reversibly, and noncompetitively inhibited by the nonsteroidal anti-inflammatory drug, niflumic acid. The active form of this powerful inhibitor (I50 = 6 X 10(-7) M) is the ionic form. We demonstrated that:
Pablo Ávalos Prado et al.
Cell, 184(2), 534-544 (2020-12-30)
Determination of what is the specificity of subunits composing a protein complex is essential when studying gene variants on human pathophysiology. The pore-forming α-subunit KCNQ1, which belongs to the voltage-gated ion channel superfamily, associates to its β-auxiliary subunit KCNE1 to
Kelly M Adams et al.
PloS one, 9(8), e103263-e103263 (2014-08-21)
The epithelium plays an active role in the response to inhaled pathogens in part by responding to signals from the immune system. Epithelial responses may include changes in chemokine expression, increased mucin production and antimicrobial peptide secretion, and changes in
Sayo Matsuba et al.
The Journal of pharmacology and experimental therapeutics, 351(3), 510-518 (2014-09-19)
The Ca(2+)-activated Cl(-) channel transmembrane proteins with unknown function 16 A (TMEM16A; also known as anoctamin 1 or discovered on gastrointestinal stromal tumor 1) plays an important role in facilitating the cell growth and metastasis of TMEM16A-expressing cancer cells. Histone
Paola Imbrici et al.
Biochimica et biophysica acta, 1838(10), 2484-2491 (2014-05-28)
CLC-K chloride channels play a crucial role in kidney physiology and genetic mutations, affecting their function are responsible for severe renal salt loss in humans. Thus, compounds that selectively bind to CLC-Ka and/or CLC-Kb channels and modulate their activity may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service