Skip to Content
Merck
All Photos(1)

Key Documents

PHR1241

Supelco

L-Methionine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-2-Amino-4-(methylmercapto)butyric acid, L-2-Amino-4-(methylthio)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
149.21
Beilstein:
1722294
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. M0960000
traceable to USP 1411504

API family

methionine & derivatives

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

284 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CSCC[C@H](N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1

InChI key

FFEARJCKVFRZRR-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

L-Methionine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by ion chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9045 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion-chromatographic determination of carbocisteine in pharmaceuticals based on non-suppressed conductimetric detection
Megoulas NC and Koupparis MA
Journal of Chromatography A, 1026(1-2), 167-174 (2004)
Ronald O Ball et al.
The Journal of nutrition, 136(6 Suppl), 1682S-1693S (2006-05-17)
Sulfur amino acid metabolism has been receiving increased attention because of the link to chronic diseases such as cardiovascular disease, Alzheimer's disease, and diabetes. In addition, the role of cysteine and optimal intakes for physiological substrates such as glutathione are
B Almé et al.
Journal of lipid research, 18(3), 339-362 (1977-05-01)
A method is described for quantitative analysis of bile acids in urine. Urine is acidified and bile acids are extracted on an Amberlite XAD-2 column. Bile salts are converted to acids on an Amberlyst A-15 column and are separated into
Olga Z Karicheva et al.
Nucleic acids research, 39(18), 8173-8186 (2011-07-05)
Mutations in human mitochondrial DNA are often associated with incurable human neuromuscular diseases. Among these mutations, an important number have been identified in tRNA genes, including 29 in the gene MT-TL1 coding for the tRNA(Leu(UUR)). The m.3243A>G mutation was described
Camilla Schalin-Jäntti et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(5), 739-747 (2013-04-05)
All patients with primary hyperparathyroidism should undergo localization studies before reoperation, but it is not known which method is most accurate. The purpose of this prospective study was to compare the performance of planar scintigraphy with (123)I/(99m)Tc-sestamibi, (99m)Tc-sestamibi SPECT (SPECT/CT)

Articles

Separation of L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material

Separation of L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material

Separation of L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material

Separation of L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service