Skip to Content
Merck
All Photos(1)

Key Documents

M4155

Sigma-Aldrich

Anti-Mouse IgG (Fab specific) antibody produced in goat

2.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Mouse Fab Antibody, Anti-Mouse Fab Antibody - Anti-Mouse IgG (Fab specific) antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

concentration

2.0 mg/mL

technique(s)

Ouchterlony double diffusion: suitable
indirect ELISA: 1:15,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG consists of four polypeptide chains, made of two identical 50 kDa γ heavy (H) chains and two identical 25 kDa κ or λ light (L) chains, which are connected by inter-chain disulfide bonds. IgG has four isotypes IgG1, IgG2, IgG3 and IgG4.

Application

Anti-Mouse IgG (Fab specific) antibody produced in goat has been used in the expression and purification of human platelet factor 4 (PF4) and anti-PF4 monoclonal antibodies.
Anti-Mouse IgG (Fab specific) antibody produced in goat was used as secondary antibody in immunocytochemical staining at a dilution of 1:10.

Biochem/physiol Actions

IgG activates the classical pathway of the complement system. It neutralizes virus particles and toxins. IgG plays a critical role in antibody-dependent cell-mediated cytotoxicity (ADCC). It has the longest serum half life and might be associated with allergy.
IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. It provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Other Notes

Antibody adsorbed with bovine, equine and human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Yuan et al.
Journal of integrative plant biology, 65(11), 2490-2504 (2023-08-07)
Soyasaponins are major small molecules that accumulate in soybean (Glycine max) seeds. Among them, type-A soyasaponins, fully acetylated at the terminal sugar of their C22 sugar chain, are responsible for the bitter taste of soybean-derived foods. However, the molecular basis
S Peller et al.
Genes, chromosomes & cancer, 21(1), 2-7 (1998-01-27)
The TP53 gene has been extensively studied in patients with chronic myeloid leukemia (CML), both in chronic phase and in blast crisis. Mutations in the gene were found in up to 30% of the patients, especially among those in blast
Qiang Sun et al.
Cell cycle (Georgetown, Tex.), 21(12), 1267-1279 (2022-03-12)
As a common cause of liver injury, hepatic ischemia/reperfusion injury (HIRI) happens in various clinical conditions including trauma, hepatectomy and liver transplantation. Since transcription factor Yin Yang 1 (YY1) was reported to be downregulated after ischemia/reperfusion (I/R) injury, we focused
Zheng Cai et al.
Nature communications, 6, 8277-8277 (2015-09-24)
Heparin-induced thrombocytopenia (HIT) is an autoimmune thrombotic disorder caused by immune complexes containing platelet factor 4 (PF4), antibodies to PF4 and heparin or cellular glycosaminoglycans (GAGs). Here we solve the crystal structures of the: (1) PF4 tetramer/fondaparinux complex, (2) PF4
Distinct Specificity and Single-molecule Kinetics of the Interaction of Pathogenic and Non-Pathogenic anti-Platelet Factor 4/Heparin Antibodies with Platelet Factor 4
Litvinov RI, et al.
The Journal of Biological Chemistry, jbc-M113 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service