Skip to Content
Merck
All Photos(3)

Documents

L7647

Sigma-Aldrich

Concanavalin A from Canavalia ensiformis (Jack bean)

Type VI, lyophilized powder

Synonym(s):

Con A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

Canavalia ensiformis

type

Type VI

form

lyophilized powder

potency

≤20 μg per mL agglutination activity

impurities

Carbohydrate, essentially free
salt, essentially free

solubility

water: 1 % (w/v)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Concanavalin A from Canavalia ensiformis (Jack bean) has been used for developing zwitterion-immobilized molecularly imprinted polymers (ziMIPs) for protein crystallization. It has been also used in the preparation of protein loaded microgels to prevent enzymatic degradation of microgels for monitoring by confocal laser scanning microscope (CLSM).

Biochem/physiol Actions

Con A is not blood group specific but has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. Ca2+ and Mn2+ ions are required for activity. Con A dissociates into dimers at pH 5.6 or below. Between pH 5.8 and pH 7.0, Con A exists as a tetramer; above pH 7.0 higher aggregates are formed. Con A exhibits mitogenic activity which is dependent on its degree of aggregation. Succinylation results in an active dimeric form which remains a dimer above pH 5.6.

Preparation Note

Highly purified by specific affinity chromatography.

Analysis Note

Agglutination activity: Determined from serial dilutions of a 1 mg/mL solution (this activity is the lowest conc. to agglutinate a 2% suspension of human erythrocytes after 1 hr incubation at 25 °C). Requires Type A Human Blood.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zwitterion-Immobilized Imprinted Polymers for Promoting the Crystallization of Proteins
Xing Y, et al.
Crystal Growth & Design, 15(10), 4932-4937 (2015)
Derivatization of epoxy-activated agarose with various carbohydrates for the preparation of stable and high-capacity affinity adsorbents: their use for affinity chromatography of carbohydrate-binding proteins.
I Matsumoto et al.
Analytical biochemistry, 116(1), 103-110 (1981-09-01)
Sevan Arabaciyan et al.
Frontiers in cell and developmental biology, 9, 607628-607628 (2021-02-16)
Single-cell variability of growth is a biological phenomenon that has attracted growing interest in recent years. Important progress has been made in the knowledge of the origin of cell-to-cell heterogeneity of growth, especially in microbial cells. To better understand the
Lindsay Kosack et al.
Scientific reports, 7(1), 11289-11289 (2017-09-14)
Lipid metabolism is increasingly being appreciated to affect immunoregulation, inflammation and pathology. In this study we found that mice infected with lymphocytic choriomeningitis virus (LCMV) exhibit global perturbations of circulating serum lipids. Mice lacking the lipid-sensing surface receptor triggering receptor
Marion Schiavone et al.
FEMS yeast research, 15(2) (2015-03-13)
The Saccharomyces cerevisiae cell surface is endowed with some relevant technological properties, notably antimicrobial and biosorption activities. For these purposes, yeasts are usually processed and packaged in an 'autolysed/dried' formula, which may have some impacts on cell surface properties. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service