Skip to Content
Merck
All Photos(1)

Key Documents

61760

Sigma-Aldrich

Lectin from Canavalia ensiformis

lyophilized, ≥50% (GE), powder

Synonym(s):

Agglutinin from jack beans, Con A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

Canavalia ensiformis

form

powder

quality

lyophilized

mol wt

Mr ~53000

concentration

≥50% (GE)

technique(s)

XRF: suitable

storage temp.

2-8°C

Biochem/physiol Actions

Con A is not blood group specific but has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. Ca2+ and Mn2+ ions are required for activity. Con A dissociates into dimers at pH 5.6 or below. Between pH 5.8 and pH 7.0, Con A exists as a tetramer; above pH 7.0 higher aggregates are formed. Con A exhibits mitogenic activity which is dependent on its degree of aggregation. Succinylation results in an active dimeric form which remains a dimer above pH 5.6.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lectins in the isolation of receptors on lymphocytes.
J J Marchalonis
Methods in enzymology, 150, 463-477 (1987-01-01)
H. Bittiger and H.P. Schnebli, eds.
Concanavalin A as a Tool, 639-639 (1976)
Dongxu Wang et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 144, 111576-111576 (2020-08-05)
Plant-derived dietary lectins have been reported to be involved in the pathogenesis of several inflammatory diseases, including hepatitis, inflammatory bowel disease, diabetes, and celiac disease. In this present study, we aimed to assess whether green tea polyphenols (GTPs) exerts protective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service