Skip to Content
Merck
All Photos(2)

Key Documents

G1145

Glass beads, acid-washed

Glass beads, acid-washed

150-212 μm (70-100 U.S. sieve)

Synonym(s):

Glass Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102422
NACRES:
NB.22
Pricing and availability is not currently available.

particle size

150-212 μm (70-100 U.S. sieve)

Related Categories

Application

Acid-washed glass beads have been used:

  • in the lysis of yeast colonies to produce a yeast two-hybrid screen[1]
  • in the extraction of DNA from Penicillium roqueforti mycelial samples[2]
  • in disruption of Streptococcus thermophilus and Lactobacillus delbrueckii subsp. bulgaricus strains for determining the enzyme activity[3]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dylan Dodd et al.
Nature, 551(7682), 648-652 (2017-11-24)
The human gut microbiota produces dozens of metabolites that accumulate in the bloodstream, where they can have systemic effects on the host. Although these small molecules commonly reach concentrations similar to those achieved by pharmaceutical agents, remarkably little is known
Timo Birkenstock et al.
The Journal of biological chemistry, 287(4), 2887-2895 (2011-12-07)
The desperate need for new therapeutics against notoriously antibiotic-resistant bacteria has led to a quest for novel antibacterial target structures and compounds. Moreover, defining targets and modes of action of new antimicrobial compounds remains a major challenge with standard technologies.
Kim I Sørensen et al.
Applied and environmental microbiology, 82(12), 3683-3692 (2016-04-24)
Streptococcus thermophilus and Lactobacillus delbrueckii subsp. bulgaricus are used in the fermentation of milk to produce yoghurt. These species normally metabolize only the glucose moiety of lactose, secreting galactose and producing lactic acid as the main metabolic end product. We
Amy Switzer et al.
Microbiology (Reading, England), 164(11), 1457-1470 (2018-06-30)
The initial adaptive transcriptional response to nitrogen (N) starvation in Escherichia coli involves large-scale alterations to the transcriptome mediated by the transcriptional activator, NtrC. One of these NtrC-activated genes is yeaG, which encodes a conserved bacterial kinase. Although it is
Fu-ping Wen et al.
Autophagy, 12(4), 671-688 (2016-04-07)
Meiosis is a special type of cellular renovation that involves 2 successive cell divisions and a single round of DNA replication. Two major degradation systems, the autophagy-lysosome and the ubiquitin-proteasome, are involved in meiosis, but their roles have yet to

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service