Skip to Content
Merck
All Photos(1)

Documents

L2879

Sigma-Aldrich

Lysozyme chloride form from chicken egg white

Grade VI, ≥35,000 units/mg protein (E1%/282)

Synonym(s):

Peptidoglycan iV-acetylmuramoyl hydrolase, Mucopeptide N-acetylmuramoylhydrolase, Muramidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

chicken egg white

type

Grade VI

form

lyophilized powder

specific activity

≥35,000 units/mg protein (E1%/282)

mol wt

~14.3 kDa

composition

Protein, ≥80%

technique(s)

cell based assay: suitable

suitability

suitable for cell lysis

application(s)

cell analysis

storage temp.

−20°C

General description

Lysozyme, also known as muramidase, is a bacteriolytic enzyme found in phagocytic cells like leukocytes. The name "lysozyme" is derived from its ability to dissolve or hydrolyze the β-1-4-glycosidic bond between N-acetylglucosamine (NAG) and N-acetylmuramic acid (NAM) in the walls of certain bacteria. This protein is present in animals, plants, bacteria, and viruses. It can also be found in granules of neutrophils, macrophages, as well as in serum, saliva, milk, honey, and hen egg white. There are different types of lysozymes, including the c-type (chicken type), the g-type (goose-type), and the i-type (invertebrates).

Application

Lysozyme chloride form from chicken egg white has been used:
  • as a component in the protein mixture to study the effects of yeast cells on membrane fouling caused by a protein mixture in dead-end filtration
  • in dip system to study its bacteriostatic or bactericidal effects on Salmonella-contaminated broiler parts
  • in viscometric assay for determining lysozyme activity    

Biochem/physiol Actions

Lysozyme serves as an indicator of macrophage-mediated host response, correlates with white cell death, and exhibits a high turnover rate. Elevated levels of serum lysozyme have been observed in various chronic inflammatory conditions, inflammatory bowel diseases, hematological disorders, and renal disorders. The c-type lysozyme from hen egg white is commonly used as a model for studying protein structure and function. Muramidase primarily exhibits bactericidal activity against Gram-positive bacteria.

Unit Definition

One unit will lyse 0.6 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 6.2 at 25 °C.

Physical form

Lyophilized powder containing sodium chloride and sodium acetate

Preparation Note

3× Crystallized

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysozyme-occurrence in nature, biological properties and possible applications
Gajda E and Bugla-P G
Journal of biological and chemical chronicles, 68, 1501-1515 (2014)
A Sobrino-López et al.
Journal of dairy science, 91(5), 1759-1768 (2008-04-19)
High-intensity pulsed electric field (HIPEF) is a nonthermal treatment studied for its wide antimicrobial spectrum on liquid food, including milk and dairy products. Moreover, the antimicrobial effect of HIPEF may be enhanced by combining HIPEF with other treatments as hurdles.
The effect of lysozyme and ethylenediaminetetraacetic acid on Salmonella on broiler parts
Samuelson, KJ, et al.
Poultry Science, 64(8), 1488-1490 (1985)
Viscometric assay for determining lysozyme activity
Yamaoka K, et al.
Analytica Chimica Acta, 219, 123-126 (1989)
R S Pappas et al.
Journal of analytical toxicology, 32(4), 281-291 (2008-04-24)
The extent to which smokeless tobacco endangers human health is an ongoing subject of debate. Studies have shown that smokeless tobacco products contain high levels of biologically available nicotine and tobacco-specific nitrosamines. Toxic metals in smokeless tobacco products have been

Protocols

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service