Skip to Content
Merck
All Photos(1)

Key Documents

L3790

Sigma-Aldrich

Lysozyme from chicken egg white

10 mg/mL

Synonym(s):

Mucopeptide N-acetylmuramoylhydrolase, Muramidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.56

biological source

chicken egg white

form

liquid

mol wt

single-chain 14.3 kDa

concentration

≥400000 units/mL
10 mg/mL

technique(s)

cell based assay: suitable

suitability

suitable for cell lysis

UniProt accession no.

application(s)

cell analysis

shipped in

wet ice

storage temp.

−20°C

Gene Information

chicken ... LYZ(396218)

Looking for similar products? Visit Product Comparison Guide

General description

Lysosyme is an enzyme found in egg white that breaks down the cell walls in bacteria enhancing the efficiency of DNA and protein extraction.

Application

Enzyme breaks down the cell walls of bacteria; used to prepare spheroplasts.
Suitable for the purification of both DNA and protein from bacteria.

Biochem/physiol Actions

Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan. Gram-negative bacteria are less susceptible due to the presence of an outer membrane and a lower proportion of peptidoglycan. However, these cells may be hydrolyzed in the presence of EDTA that chelates metal ions in the outer bacterial membrane.

The enzyme is active over a broad pH range (6.0 to 9.0). At pH 6.2, maximal activity is observed over a wider range of ionic strengths (0.02 to 0.100 M) than at pH 9.2 (0.01 to 0.06 M).
Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan.

Suitability

Tested for suitability with CelLytic B in protein extraction from bacteria.

Unit Definition

One unit will lyse 0.4 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 8.0 at 37 °C.

Physical form

Ready to use solution:10 mg/mL lysozyme in 25 mM sodium acetate pH 4.5 with 50% glycerol.
Solution in 50% glycerol, 25 mM Sodium Acetate

Legal Information

CelLytic is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase-sensitive spatially-modulated surface plasmon resonance polarimetry for detection of biomolecular interactions
Patskovsky,S et al.
Sens. Actuators, 133 (2), 628-631 (2008)
Daimon P Simmons et al.
Journal of immunology (Baltimore, Md. : 1950), 185(4), 2405-2415 (2010-07-28)
Dendritic cells (DCs) cross process exogenous Ags and present them by class I MHC (MHC-I) molecules to CD8(+) T cells specific for Ags from viruses and bacteria such as Mycobacterium tuberculosis. Unmethylated CpG DNA signals through TLR9 to induce type
Michael G Drage et al.
Cellular immunology, 258(1), 29-37 (2009-04-14)
Mycobacterium tuberculosis (Mtb) signals through Toll-like receptor 2 (TLR2) to regulate antigen presenting cells (APCs). Mtb lipoproteins, including LpqH, LprA, LprG and PhoS1, are TLR2 agonists, but their co-receptor requirements are unknown. We studied Mtb lipoprotein-induced responses in TLR2(-/-), TLR1(-/-)
Benjamin S Waldman et al.
Cell, 180(2), 359-372 (2020-01-21)
Toxoplasma gondii chronically infects a quarter of the world's population, and its recrudescence can cause life-threatening disease in immunocompromised individuals and recurrent ocular lesions in the immunocompetent. Acute-stage tachyzoites differentiate into chronic-stage bradyzoites, which form intracellular cysts resistant to immune
P F Zabrodskiĭ et al.
Eksperimental'naia i klinicheskaia farmakologiia, 75(11), 19-21 (2013-01-18)
It is established in experiments on noninbred rats that the use of imunofan (20 mg/kg daily) and polyoxidonium (150 mg/kg daily) for 7 days on the background of chronic intoxication with organophosphorus agent VX (0.01 LD50, single daily treatment for

Protocols

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service