Skip to Content
Merck
All Photos(4)

Documents

M3770

Sigma-Aldrich

Micrococcus lysodeikticus ATCC No. 4698

suitable for substrate for the assay of lysozyme, lyophilized cells

Synonym(s):

Micrococcus luetus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized cells

Quality Level

suitability

suitable for substrate for the assay of lysozyme

storage temp.

−20°C

Application

Micrococcus lysodeikticus ATCC No. 4698 has been used in a study to assess lysozyme separation by hollow-fibre ultrafiltration. It has also been used in a study to investigate the encapsulation of protein drugs in biodegradable microparticles.
Lysozyme lysates harvested from cultures of Micrococcus lysodeikticus were attached to sepharose and used for affinity chromatography to isolate various bacteriolytic enzymes.

Biochem/physiol Actions

Micrococcus luetus is a Gram-positive bacteria that is identified by the release of yellow water-insoluble pigments. This species requires succinic acid for its growth and is found to be susceptible to β-lytic metalloendopeptidase lyses by Lysobacter enzymogenes. Its membrane includes enzymes that participate in the prenylation reactions by utilizing prenyl pyrophosphates as donors. M. luteus is known to be used for cloning the cis-prenyl transferase gene.

Quality

Contains polynucleotide phosphorylase.

Unit Definition

One unit will lyse 0.6 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 6.2 at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia C Sánchez et al.
BMC genomics, 12, 626-626 (2011-12-23)
Fish under intensive culture conditions are exposed to a variety of acute and chronic stressors, including high rearing densities, sub-optimal water quality, and severe thermal fluctuations. Such stressors are inherent in aquaculture production and can induce physiological responses with adverse
Tania Jauslin et al.
mBio, 12(1) (2021-02-18)
Ingestion and killing of bacteria by phagocytic cells protect the human body against infections. While many mechanisms have been proposed to account for bacterial killing in phagosomes, their relative importance, redundancy, and specificity remain unclear. In this study, we used
Ingrid Bourgeois et al.
FEMS microbiology letters, 290(1), 105-113 (2008-11-26)
The nucleotide sequence of atlL, a gene encoding a putative Staphylococcus lugdunensis peptidoglycan hydrolase, was determined using degenerate consensus PCR and genome walking. This 3837-bp gene encodes a protein, AtlL, that appears as a putative bifunctional autolysin with a 29-amino
Jingwei Xie et al.
Journal of colloid and interface science, 317(2), 469-476 (2007-10-20)
A co-axial electrospray process was developed to encapsulate protein-based drugs in biodegradable polymeric microparticles eliminating the key problem faced by other conventional methods of protein encapsulation--the primary emulsion being a major cause for protein denaturation and aggregation. Bovine serum albumin
G L Lorca et al.
Current microbiology, 42(1), 39-44 (2000-12-16)
Antibacterial activity of 17 strains of lactobacilli was tested against 10 strains of H. pylori. The inhibition observed was related to the acid production and the low pH attained. No relationship between CagA phenotype of H. pylori strains and tolerance

Protocols

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service