Skip to Content
Merck
All Photos(1)

Documents

I0503

Sigma-Aldrich

Isopentenyl pyrophosphate triammonium salt solution

1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)

Synonym(s):

IPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O7P2 · 3NH3
CAS Number:
Molecular Weight:
297.18
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (TLC)

packaging

vial of 200 μg

concentration

1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3))

storage temp.

−20°C

SMILES string

N.N.N.CC(=C)CCOP(O)(=O)OP(O)(O)=O

InChI

1S/C5H12O7P2.3H3N/c1-5(2)3-4-11-14(9,10)12-13(6,7)8;;;/h1,3-4H2,2H3,(H,9,10)(H2,6,7,8);3*1H3

InChI key

JREYOWJEWZVAOR-UHFFFAOYSA-N

Application

Intermediate in terpene biosynthesis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karan Malhotra et al.
Molecular plant, 9(11), 1464-1477 (2016-10-25)
Artemisinin is highly effective against drug-resistant malarial parasites, which affects nearly half of the global population and kills >500 000 people each year. The primary cost of artemisinin is the very expensive process used to extract and purify the drug from Artemisia
Jarika Kaewbanjong et al.
The Journal of pharmacy and pharmacology, 69(12), 1716-1723 (2017-08-25)
To investigate the efficacy of clotrimazole microemulsion (CTZ-ME) and its gel form, clotrimazole microemulsion-based gel (CTZ-MBG), for the treatment of oral candidiasis. CTZ-ME and CTZ-MBG were characterized for droplet size and texture, respectively. The ex-vivo permeation study and irritancy assessment
Zrinka Karačić et al.
Biological chemistry, 398(1), 101-112 (2016-07-29)
In a search for plant homologues of dipeptidyl peptidase III (DPP III) family, we found a predicted protein from the moss Physcomitrella patens (UniProt entry: A9TLP4), which shared 61% sequence identity with the Arabidopsis thaliana uncharacterized protein, designated Nudix hydrolase
John M Lacy et al.
Urology, 98, 189-194 (2016-07-05)
To identify comorbid conditions that may increase the likelihood of revision surgery after primary penile prosthesis implantation. To evaluate trends in utilization of prostheses and selection of device within the Veterans Affairs (VA) system. A retrospective review of the VA
Alanna S Murday et al.
Immunology, 151(4), 385-394 (2017-03-28)
The study aimed to identify an immunoregulatory factor that restores the phosphoantigen response of Vγ9Vδ2

Articles

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service