Skip to Content
Merck
All Photos(1)

Key Documents

76532

Sigma-Aldrich

Geranyl pyrophosphate lithium salt

≥95.0% (TLC)

Synonym(s):

(E)-3,7-Dimethyl-2,6-octadien-1-ol trihydrogen pyrophosphate lithium salt, (E)-3,7-Dimethyl-2,6-octadien-1-yl pyrophosphate lithium salt, GPP-Li

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O7P2 · xLi+
CAS Number:
Molecular Weight:
314.21 (free acid basis)
Beilstein:
1915690
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.25
Assay:
≥95.0% (TLC)
form:
powder

Quality Level

Assay

≥95.0% (TLC)

form

powder

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite in monoterpenoid biosynthesis, substrate for monoterpene synthases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Tringali et al.
Neuroscience letters, 354(2), 107-110 (2003-12-31)
Statins were recently shown to possess anti-inflammatory activities, which might be responsible for their favourable effects in cardiovascular or CNS disorders independently from the inhibition of hydroxy-methyl-glutaryl CoA reductase. Here we investigated the effects of the statins lovastatin and mevastatin
Fu-Lien Hsieh et al.
Journal of molecular biology, 404(5), 859-873 (2010-10-23)
Isoprenoids, most of them synthesized by prenyltransferases (PTSs), are a class of important biologically active compounds with diverse functions. The mint geranyl pyrophosphate synthase (GPPS) is a heterotetramer composed of two LSU·SSU (large/small subunit) dimers. In addition to C(10)-GPP, the
Annie P-C Chen et al.
The Biochemical journal, 386(Pt 1), 169-176 (2004-09-28)
UPPS (undecaprenyl pyrophosphate synthase) catalyses consecutive condensation reactions of FPP (farnesyl pyrophosphate) with eight isopentenyl pyrophosphates to generate C55 UPP, which serves as a lipid carrier for bacterial peptidoglycan biosynthesis. We reported the co-crystal structure of Escherichia coli UPPS in
F Pont et al.
Analytical chemistry, 73(15), 3562-3569 (2001-08-21)
New phosphorylated microbial metabolites referred to as phosphoantigens activate immune responses in humans. Although these molecules have leading applications in medical research, no direct method allows their rapid and unambiguous structural identification. Here, we interfaced online HPAEC (high performance anion-exchange
V G Ladygin
Zhurnal obshchei biologii, 63(4), 299-325 (2002-09-27)
The author discusses the current state of biochemical and genetic aspects of carotenoid biosynthesis in chloroplasts of algae and higher plants. Two ways of biosynthesis of key C5-isoperene units have been considered: 1) from acetate (C2) via mevalonic acid (C6)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service