Skip to Content
Merck
All Photos(1)

Key Documents

D5794

Sigma-Aldrich

Diacylglycerol Kinase Inhibitor II

solid

Synonym(s):

3-[2-[4-(bis(4-Fluorophenyl)methylene)-1-piperidinyl]ethyl]-2,3-dihydro-2-thioxo-4(1H)-quinazolinone, R59949

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H25F2N3OS
CAS Number:
Molecular Weight:
489.58
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥97% (HPLC)

form

solid

color

pale yellow

mp

228-230  °C

solubility

0.1 M HCl: slightly soluble
0.1 M NaOH: slightly soluble
DMSO: soluble
H2O: insoluble
ethanol: soluble
ethyl acetate: soluble

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)\C(=C2\CCN(CCN3C(=S)Nc4ccccc4C3=O)CC2)c5ccc(F)cc5

InChI

1S/C28H25F2N3OS/c29-22-9-5-19(6-10-22)26(20-7-11-23(30)12-8-20)21-13-15-32(16-14-21)17-18-33-27(34)24-3-1-2-4-25(24)31-28(33)35/h1-12H,13-18H2,(H,31,35)

InChI key

ZCNBZFRECRPCKU-UHFFFAOYSA-N

Related Categories

Application

Diacylglycerol Kinase Inhibitor II has been used to determine tumor-induced inhibition with genetically modified cytotoxic T cells expressing chimeric antigen receptors (CAR). It has also been used to induce pAkt and PKR-like extracellular signal-regulated kinase (pErk) signals in T-cell acute lymphoblastic leukemia (T-ALL) cells.

Biochem/physiol Actions

Diacylglycerol kinase inhibitor. Inhibits formation of [38P]1-Oleoyl-2-acetylglyceryl-3-phosphoric acid (OAPA) in red blood cell membranes: IC50 = 3.3 μM.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marylana Saadeh et al.
PloS one, 7(1), e30200-e30200 (2012-01-25)
Chronic exposure (24-72 hrs) of pancreatic islets to elevated glucose and fatty acid leads to glucolipoxicity characterized by basal insulin hypersecretion and impaired glucose-stimulated insulin secretion (GSIS). Our aim was to determine the mechanism for basal hypersecretion of insulin. We
Costin N Antonescu et al.
Molecular biology of the cell, 21(16), 2944-2952 (2010-06-25)
Clathrin-mediated endocytosis (CME) is the main route of internalization of receptor-ligand complexes. Relatively little is known about the role of specific lipids in CME, in particular that of phosphatidic acid (PA). We examined the effect of altering cellular PA levels
RasGRP1 overexpression in T-ALL increases basal nucleotide exchange on Ras rendering the Ras/PI3K/Akt pathway responsive to protumorigenic cytokines
Ksionda O, et al.
Oncogene, 35(28), 3658-3658 (2016)
Elisa Temes et al.
The Journal of biological chemistry, 280(25), 24238-24244 (2005-04-26)
Hypoxia-inducible factors (HIF) are heterodimeric (alpha/beta) transcription factors that play a fundamental role in cellular adaptation to low oxygen tension. In the presence of oxygen, the HIF-alpha subunit becomes hydroxylated at specific prolyl residues by prolyl hydroxylases. This post-translational modification
R W Erickson et al.
The Journal of biological chemistry, 274(32), 22243-22250 (1999-07-31)
The superoxide-generating neutrophil NADPH oxidase can be activated in cell-free reconstitution systems by several agonists, most notably arachidonic acid and the detergent sodium dodecyl sulfate. In this study, we show that both phosphatidic acids and diacylglycerols can serve separately as

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service