Skip to Content
Merck

Skip To

95098

(E)-1-Hydroxy-2-methyl-2-butenyl 4-pyrophosphate lithium salt

≥95.0% (TLC)

Synonym(s):

(E)-4-Hydroxy-3-methylbut-2-enyl diphosphate lithium salt, 1-Hydroxy-2-methyl-2-(E)-butenyl 4-diphosphate, HDMAPP, HMBPP, HO-DMAPP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H12O8P2 · xLi+
CAS Number:
Molecular Weight:
262.09 (free acid basis)
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.85
Assay:
≥95.0% (TLC)
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (TLC)

form

solid

storage temp.

−20°C

SMILES string

[P](=O)([O-])([O-])O[P](=O)([O-])OCC=C(CO)C

InChI

1S/C5H12O8P2/c1-5(4-6)2-3-12-15(10,11)13-14(7,8)9/h2,6H,3-4H2,1H3,(H,10,11)(H2,7,8,9)/p-3

InChI key

MDSIZRKJVDMQOQ-UHFFFAOYSA-K

Biochem/physiol Actions

HMB-PP is an intermediate of the non-mevalonate pathway (MEP pathway) of isoprenoid biosynthesis[1][2]; it was shown to be a potent γδ T cell activator[3][4].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
374429659956901
form

solid

form

powder

form

solid

form

-

assay

≥95.0% (TLC)

assay

≥95.0% (TLC)

assay

≥90.0% (TLC)

assay

≥95.0% (TLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. I would like to know in which liquid should the HMBPP be resuspended, please?

    1 answer
    1. This product can be dissolved in water.

      Helpful?

Reviews

No rating value

Active Filters