Skip to Content
Merck
All Photos(4)

Documents

H3663

Sigma-Aldrich

Anti-HA antibody, Mouse monoclonal

clone HA-7, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-HA, Anti-HA, Anti-Influenza Hemagglutinin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HA-7, monoclonal

concentration

~1 mg/mL

technique(s)

immunocytochemistry: 1.0-2.0 μg/mL
immunoprecipitation (IP): 0.5-2.0 μg
western blot: 0.25-1 μg/mL

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Hemagglutinin (HA) is a glycoprotein present in the membrane of influenza virus. HA exists as a trimer and each monomer at its membrane tip has receptor-binding site. Glycosylation event of HA occurs in endoplasmic reticulum. The head region with the receptor-binding site of the HA is variable. The mouse monoclonal Anti-HA, Clone HA-7 recognizes the human influenza virus hemagglutinin (HA), known as the HA tag. The product is reactive with HA-tagged fusion proteins expressed at either the amino or the carboxy terminus of the fusion protein.

Specificity

The antibody recognizes native as well as denatured-reduced forms of HA-tagged proteins and is reactive with N- or C-terminal HA-tagged fusion proteins expressed in E. coli or in mammalian cells.

Immunogen

synthetic peptide corresponding to a fragment of human influenza virus hemagglutinin (HA) known as HA-tag, conjugated to KLH

Application

Monoclonal Anti-HA antibody produced in mouse has been used:
  • in immunoblotting
  • in immunocytochemistry
  • in the immunoprecipitation

Biochem/physiol Actions

Hemagglutinin (HA) binds to acid binding and is essential for the initiation of virus infection. It is the key target for the generation of vaccine. HA is the dominant antigen against which humoral immune response is provoked. Vaccines development involves the generation of antibody specific to the variable head and the conserved stalk region. Both the anti-stalk and anti-head antibodies may elicit protective function in influenza disease.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide as a preservative.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ribosomal protein L4 interacts with viral protein VP3 and regulates the replication of infectious bursal disease virus
Chen Y, et al.
Virus Research, 211, 73-78 (2016)
Yumei Zheng et al.
Nature communications, 10(1), 3600-3600 (2019-08-11)
Autophagy depends on the E2 enzyme, Atg3, functioning in a conserved E1-E2-E3 trienzyme cascade that catalyzes lipidation of Atg8-family ubiquitin-like proteins (UBLs). Molecular mechanisms underlying Atg8 lipidation remain poorly understood despite association of Atg3, the E1 Atg7, and the composite
Influenza hemagglutinin and neuraminidase membrane glycoproteins
Gamblin SJ and Skehel JJ
The Journal of Biological Chemistry, 285(37), 28403-28409 (2010)
Evaluation of pre-existing anti-hemagglutinin stalk antibody as a correlate of protection in a healthy volunteer challenge with influenza A/H1N1pdm virus
Park JK, et al.
mBio, 9(1), e02284-e02217 (2018)
A newly cloned ClC-3 isoform, ClC-3d, as well as ClC-3a mediates Cd2+-sensitive outwardly rectifying anion currents
Okada T, et al.
Cellular Physiology and Biochemistry, 33(3), 539-556 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service