H3265
Methyl heneicosanoate
~99% (capillary GC), crystalline
Synonym(s):
Heneicosanoic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)19COOCH3
CAS Number:
Molecular Weight:
340.58
Beilstein:
1792879
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
~99% (capillary GC)
Quality Level
form
crystalline
bp
207 °C/4 mmHg (lit.)
mp
48-50 °C (lit.)
functional group
ester
lipid type
saturated FAs
shipped in
ambient
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCCCCCCC(=O)OC
InChI
1S/C22H44O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24-2/h3-21H2,1-2H3
InChI key
AJRICDSAJQHDSD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113.00 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
K D Cox et al.
Mycological research, 110(Pt 4), 414-422 (2006-03-21)
Limited information is available regarding the composition of cellular fatty acids in Armillaria and the extent to which fatty acid profiles can be used to characterize species in this genus. Fatty acid methyl ester (FAME) profiles generated from cultures of
R J Horvat
Poultry science, 57(3), 827-828 (1978-05-01)
Lipids from the skin of commercially processed broilers were extracted by Folch's technique and analyzed for fatty acids as their methyl esters by gas-liquid chromatography/mass pectrometry. Of the 24 fatty acids, identified, 5 had not been previously identified in either
Yoshinori Masukawa et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 823(2), 131-142 (2005-07-12)
A systematic method for the sensitive, precise and accurate determination of hair lipids, including trace amounts of intrinsic endogenous cholesterol (CH), ceramide/N-palmitoyl-DL-dihydrosphingosine (CER/PDS), cholesterol sulfate (CS) and chemically bound 18-methyl eicosanoic acid (18-MEA), has been developed in combination with TLC/FID
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service