1.15186
Hydrochloric acid
36%, SUPRAPUR®
Synonym(s):
Hydrochloric acid
About This Item
Recommended Products
Product Name
Hydrochloric acid, Suprapur®, 36%
Agency
suitable for GB 5009.253-2016
suitable for SM 5210
Quality Level
vapor density
1.3 (vs air)
vapor pressure
190 hPa ( 20 °C)
3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)
description
for inorganic trace analysis
Assay
36%
form
liquid
pH
<1 (20 °C in H2O)
bp
>100 °C (lit.)
density
1.18 g/L at 20 °C
1.18 g/cm3 at 20 °C
storage temp.
15-25°C
SMILES string
Cl
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.
Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.
Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.
Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.
Related Content
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service