Skip to Content
Merck
All Photos(3)

Documents

D0627

Sigma-Aldrich

N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt

≥96% (HPLC), powder

Synonym(s):

Bucladesine sodium salt, Dibutyryl cAMP sodium salt, Dibutyryl cyclic-AMP sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H23N5O8PNa
CAS Number:
Molecular Weight:
491.37
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥96% (HPLC)

form

powder

impurities

≤0.5% Cyclic AMP
≤3% monobutyryl derivatives

color

off-white

solubility

H2O: 100 mg/mL

storage temp.

−20°C

SMILES string

[Na+].CCCC(=O)Nc1ncnc2n(cnc12)[C@@H]3O[C@@H]4COP([O-])(=O)O[C@H]4[C@H]3OC(=O)CCC

InChI

1S/C18H24N5O8P.Na/c1-3-5-11(24)22-16-13-17(20-8-19-16)23(9-21-13)18-15(30-12(25)6-4-2)14-10(29-18)7-28-32(26,27)31-14;/h8-10,14-15,18H,3-7H2,1-2H3,(H,26,27)(H,19,20,22,24);/q;+1/p-1/t10-,14-,15-,18-;/m1./s1

InChI key

KRBZRVBLIUDQNG-JBVYASIDSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Dibutyryl CAMP is a cell-permeable CAMP analogue that activates CAMP dependent protein kinase (PKA) or the CAMP/PKA signaling pathway. Dibutyryl CAMP is used as a morphological differentiation inducer and cell signaling modulator in cells such as Schwann cells. Dibutyrl cAMP is used in a variety of differentiation studies, such as neuron differentiation / neuronal differentiation.

Biochem/physiol Actions

Cell-permeable cAMP analog that activates cAMP dependent protein kinase (PKA).

Caution

Loses 2′-O-butyryl group at pH 8.5

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junji Yamauchi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(35), 12579-12592 (2011-09-02)
In development of the peripheral nervous system, Schwann cells proliferate, migrate, and ultimately differentiate to form myelin sheath. In all of the myelination stages, Schwann cells continuously undergo morphological changes; however, little is known about their underlying molecular mechanisms. We
De-Yi Liu et al.
Fertility and sterility, 99(1), 86-91 (2012-09-19)
To determine association between defective protein kinases C (PKC) and A (PKA) and disordered zona pellucida (ZP)-induced acrosome reaction (DZPIAR) in normozoospermic infertile men with normal sperm-ZP binding. Sperm from DZPIAR infertile men were treated without (control) or with (test)
Annelieke S Wentzel et al.
Scientific reports, 10(1), 13470-13470 (2020-08-12)
Mammalian macrophages can adopt polarization states that, depending on the exact stimuli present in their extracellular environment, can lead to very different functions. Although these different polarization states have been shown primarily for macrophages of humans and mice, it is
Steven L Coon et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(33), 13319-13324 (2012-08-07)
Long noncoding RNAs (lncRNAs) play a broad range of biological roles, including regulation of expression of genes and chromosomes. Here, we present evidence that lncRNAs are involved in vertebrate circadian biology. Differential night/day expression of 112 lncRNAs (0.3 to >50
Adam Wieschhaus et al.
The Biochemical journal, 448(1), 141-152 (2012-08-09)
Pharmacological inhibitors of cysteine proteases have provided useful insights into the regulation of calpain activity in erythrocytes. However, the precise biological function of calpain activity in erythrocytes remains poorly understood. Erythrocytes express calpain-1, an isoform regulated by calpastatin, the endogenous

Protocols

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service