Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0618-05

Protein G Sepharose 4 Fast Flow

Cytiva 17-0618-05, pack of 200 mL

Synonym(s):

Fast Flow resin, Antibody purification resin, IgG purification resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

recombinant protein G lacking albumin-binding region

packaging

pack of 200 mL

manufacturer/tradename

Cytiva 17-0618-05

storage condition

(20% Ehtanol)

matrix

4% cross-linked agarose

average diameter

90 μm (d50v)

cleaning in place

2-10

working range

3-9

suitability

suitable for bioprocess medium

storage temp.

2-8°C

General description

Protein G Sepharose 4 Fast Flow is recombinant protein G coupled to Sepharose 4 Fast Flow.

Protein G Sepharose 4 Fast Flow has recombinant protein G immobilized by the cyanogen bromide (CNBr) method to Sepharose 4 Fast Flow. Protein G exhibit binding specificities that complement Protein A media and binds to the Fc region of IgG from a variety of mammalian species. Protein G Sepharose 4 Fast Flow may be used to isolate and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.

As member of the BioProcess media range, Protein G Sepharose 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • Binding specificities that complement Protein A media.
  • Binds a broad range of IgG species and subclasses.
  • Multi-point attachment minimizes ligand leakage.
  • Used in a range of research applications.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monique G P van der Wijst et al.
bioRxiv : the preprint server for biology (2021-03-25)
Type I interferon (IFN-I) neutralizing autoantibodies have been found in some critical COVID-19 patients; however, their prevalence and longitudinal dynamics across the disease severity scale, and functional effects on circulating leukocytes remain unknown. Here, in 284 COVID-19 patients, we found
Kanika Vanshylla et al.
Cell host & microbe, 29(6), 917-929 (2021-05-14)
Understanding antibody-based SARS-CoV-2 immunity is critical for overcoming the COVID-19 pandemic and informing vaccination strategies. We evaluated SARS-CoV-2 antibody dynamics over 10 months in 963 individuals who predominantly experienced mild COVID-19. Investigating 2,146 samples, we initially detected SARS-CoV-2 antibodies in 94.4%
Neutralizing Autoantibodies to Type I Interferons in COVID-19 Convalescent Donor Plasma.
Sara E Vazquez et al.
Journal of clinical immunology, 41(6), 1169-1171 (2021-05-20)
Kanika Vanshylla et al.
Cell host & microbe, 30(1), 69-82 (2022-01-02)
A fraction of COVID-19 convalescent individuals mount a potent antibody response to SARS-CoV-2 with cross-reactivity to SARS-CoV-1. To uncover their humoral response in detail, we performed single B cell analysis from 10 SARS-CoV-2 elite neutralizers. We isolated and analyzed 126
Monique G P van der Wijst et al.
Science translational medicine, 13(612), eabh2624-eabh2624 (2021-08-26)
Neutralizing autoantibodies against type I interferons (IFNs) have been found in some patients with critical coronavirus disease 2019 (COVID-19), the disease caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). However, the prevalence of these antibodies, their longitudinal dynamics across

Articles

Purify monoclonal or polyclonal IgG from serum, cell culture supernatant or ascitic fluid using the HiTrap Protein G HP from Cytiva, an affinity-exclusion chromatography product containing Sepharose-immobilized Protein G.

This page describes immunoprecipitation (immunoaffinity or pull-down techniques).

This page describes efficient column packing and preparation for affinity chromatography of antibodies.

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

See All

Protocols

This page provides information about different pull-down assays for the further isolation of multiprotein complexes to identify their components with products from Cytiva.

This page shows how to separate IgG antibodies by affinity chromatography using Protein G Sepharose 4 Fast Flow from Cytiva.

This page shows how to prepare samples for purification with affinity chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service