Skip to Content
Merck
All Photos(1)

Documents

G5262

Supelco

GAPDH

standard for protein electrophoresis

Synonym(s):

Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, D-Glyceraldehyde 3-phosphate:NAD+ oxidoreductase (phosphorylating), GAPDH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

grade

for molecular biology

Quality Level

form

powder

mol wt

~36 kDa

packaging

vial of 5 mg

technique(s)

electrophoresis: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

GAPDH (Glyceraldehyde-3-phosphate dehydrogenase) catalyzes the conversion of glyceraldehyde-3-phosphate into D-glycerate-1,3-bisphosphate as part of the glycolysis pathway. GAPDH has also been found to function in additional cellular process, such as transcription, apoptosis, oxidative stress and ER to Golgi transport.

Application

GAPDH protein is suitable for use as a molecular weight marker and protein standard for molecular biology applications, including western blotting and mass spectometry.

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase catalyzes the conversion of glyceraldehyde-3-phosphate into D-glycerate-1,3-bisphosphate as part of the glycolysis pathway.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling Ji et al.
Molecular therapy. Nucleic acids, 19, 546-561 (2020-01-14)
Long non-coding RNAs (lncRNAs) are crucial molecules in tumorigenesis and tumor growth in various human cancers, including colorectal cancer (CRC). Studies have revealed that lncRNAs can regulate cellular processes in cancers by interacting with proteins, for example RNA-binding proteins (RBPs).
Kati Juuti-Uusitalo et al.
Investigative ophthalmology & visual science, 54(5), 3510-3519 (2013-05-21)
Aquaporins (AQPs), a family of transmembrane water channel proteins, are essential for allowing passive water transport through retinal pigmented epithelial (RPE) cells. Even though human native RPE cells and immortalized human RPEs have been shown to express AQPs, the expression
Xiaopeng Wang et al.
Molecular medicine reports, 23(2) (2020-12-11)
Acute progressive hypoxic respiratory failure caused by various predisposing factors is known as acute respiratory distress syndrome (ARDS). Although penehyclidine hydrochloride (PHC), an anticholinergic drug, is widely applied in clinical practice, the specific mechanisms underlying PHC in the treatment of
Craig Nicholls et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(33), 13308-13313 (2012-08-01)
Oxidative stress regulates telomere homeostasis and cellular aging by unclear mechanisms. Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) is a key mediator of many oxidative stress responses, involving GAPDH nuclear translocation and induction of cell death. We report here that GAPDH interacts with
J E Welch et al.
Journal of andrology, 21(2), 328-338 (2000-03-14)
Although the process of glycolysis is highly conserved in eukaryotes, several glycolytic enzymes have unique structural or functional features in spermatogenic cells. We previously identified and characterized the mouse complementary DNA (cDNA) and a gene for 1 of these enzymes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service