Skip to Content
Merck
All Photos(1)

Key Documents

GERPN2209

ECL Western Blotting Detection Reagents

Cytiva RPN2209

Synonym(s):

ECL detection reagents, Western blot reagents

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105332
NACRES:
NA.32

feature

wetted part: no

packaging

pkg of 1 kit

manufacturer/tradename

Cytiva RPN2209

color

Detection reagent 1: Colorless Detection reagent 2: Colorless

storage temp.

2-8°C

General description

Amersham ECL Western Blotting Detection Reagents were the world′s first commercial chemiluminescent reagents for Western blotting and are still the most widely used. They are at least 10-fold more sensitive than colorimetric assays and produce excellent and reliable results every time.

These reagents use enhanced luminol-based detection which is suitable for all routine western blotting. ECL Western Blotting Detection Reagents in combination with Hybond® ECL membranes form a complete solution for Western blot applications.

Contents include the following:
GERPN2109, 62.5 mL each of ECL Detection Reagents 1 and 2 (1000 cm2)
GERPN2209, 125 mL each (2000 cm2)
GERPN2106, 250 mL each (4000 cm2)
GERPN2134, 375 mL each (6000 cm2)
Emission duration max. 2 h

Application

Western Blotting

Features and Benefits

  • High sensitivity, ~10 pg in model system.
  • Proven performance, the most frequently cited ECL Western Blotting Detection Reagents in publications
  • Detectable on Hyperfilm ECL or with CCD imager
  • Recommended mebrane Hybond ECL

Components

Sufficient for detection of 2000 cm2 of membrane:
125 mL Detection reagent 1(color : colorless)
125 mL Detection reagent 2 (color: colorless)

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Amersham is a trademark of Cytiva
ECL is a trademark of Cytiva
Hybond is a registered trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Zhang et al.
International immunopharmacology, 69, 88-94 (2019-01-28)
Inflammation is fundamental in osteoarthritis (OA) pathogenesis. Semaphorin 4A (Sema4A) has been implicated in immune-associated diseases, however, its role in OA remains unclear. In this study, we show that Sema4A is upregulated in knee OA articular cartilage as well as
Qinmiao Huang et al.
Molecular medicine reports, 16(5), 7086-7091 (2017-09-14)
Lung cancer is reported to be a major public health issue worldwide and the overall prognosis of patients remains poor. The expression levels of Livin and Survivin, of the inhibitors of apoptosis (IAP) family, are associated with prognostic significance in
Gennaro Gambardella et al.
Science advances, 6(39) (2020-09-27)
Cells respond to starvation by shutting down protein synthesis and by activating catabolic processes, including autophagy, to recycle nutrients. This two-pronged response is mediated by the integrated stress response (ISR) through phosphorylation of eIF2α, which represses protein translation, and by
Ping Zhou et al.
International journal of biological sciences, 16(2), 204-215 (2020-01-14)
Chronic renal failure (CRF), also known as chronic kidney disease (CKD), is a common renal disorder characterized by gradual kidney dysfunction. Molecular dissection reveals that transforming growth factor beta (TGF-β) plays a central role in the pathogenesis of CRF. However

Articles

Background and protocols describing the various methods used by molecular biologists to detect samples of protein or nucleic acids bound to membranes.

This page describes troubleshooting strategies in the detection of GST-tagged proteins.

Protocols

This page describes chemiluminescence detection of GST-tagged proteins with Amersham ECL, Amersham ECL Prime, and Amersham ECL Select from Cytiva.

This page describes how to monitor expression and purification of GST-tagged proteins by Western blot analysis using Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service