Skip to Content
Merck
All Photos(1)

Key Documents

G6019

Sigma-Aldrich

Glyceraldehyde-3-phosphate Dehydrogenase from human erythrocytes

lyophilized powder, 50-150 units/mg protein

Synonym(s):

D-Glyceraldehyde 3-phosphate:NAD+ oxidoreductase (phosphorylating), GAPDH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

specific activity

50-150 units/mg protein

composition

Protein, 40-70% biuret

UniProt accession no.

foreign activity

3-Phosphoglyceric phosphokinase ≤2%

storage temp.

−20°C

Gene Information

human ... GAPDH(2597)

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase catalyzes the conversion of glyceraldehyde-3-phosphate to 1,3-bisphosphoglycerate as part of glycolysis. It has also been shown to have roles in initiation of apoptosis, transcription activation and the shuttling of ER to Golgi vesicles<<<18,New>>>.

Unit Definition

One unit will reduce 1.0 μmole of 3-phosphoglycerate to D-glyceraldehyde 3-phosphate per min in a coupled system with 3-phosphoglyceric phosphokinase at pH 7.6 at 25 °C.

Physical form

Lyophilized powder containing sodium citrate buffer salts

inhibitor

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Salemi et al.
Tumori, 98(3), 86e-88e (2012-07-25)
Head and neck squamous cell carcinoma (HNSCC) is one of the most common cancers in the world. Risk factors for this cancer include tobacco and alcohol use, ultraviolet light exposure, and viral infection. Parkinson's disease is one of the most
Anne Gürtler et al.
Analytical biochemistry, 433(2), 105-111 (2012-10-23)
Western blots are used to specifically measure the relative quantities of proteins of interest in complex biological samples. Quantitative measurements can be subject to error due to process inconsistencies such as uneven protein transfer to the membrane. These non-sample-related variations
Kyle L Sunderland et al.
Journal of strength and conditioning research, 27(1), 1-7 (2012-10-23)
The purpose of this study was to investigate how age and 1 week of conventional resistance exercise affects commonly used housekeeping gene (HKG) messenger RNAs (mRNAs) in skeletal muscle. Ten college-aged (18-25 years) and 10 older (60-76 years) men completed
C Fossum et al.
Veterinary immunology and immunopathology, 150(3-4), 141-148 (2012-10-06)
The expression of tlr4, md2 and cd14 was studied in equine blood leukocytes and in intestinal samples using real time PCR. The stability of three commonly used reference genes, glyceraldehyde-3P-dehydrogenase (GAPDH), hypoxantine ribosyltransferase (HPRT) and succinate dehydrogenase complex subunit A
Craig Nicholls et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(33), 13308-13313 (2012-08-01)
Oxidative stress regulates telomere homeostasis and cellular aging by unclear mechanisms. Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) is a key mediator of many oxidative stress responses, involving GAPDH nuclear translocation and induction of cell death. We report here that GAPDH interacts with

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service