Skip to Content
Merck
All Photos(1)

Key Documents

C1992

Sigma-Aldrich

ColorBurst Electrophoresis Marker

mol wt 8,000-220,000 Da

Synonym(s):

protein markers, protein molecular weight markers, protein standards

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105337
NACRES:
NA.25
Pricing and availability is not currently available.

form

solution

Quality Level

mol wt

8,000-220,000 Da

pH

8

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Brilliantly colored, exceptionally well-resolved, convenient, and stable, ColorBurst protein molecular weight markers perform impressively in a variety of gel compositions and concentrations. ColorBurst Markers are composed of 8 polypeptides which have been chemically reduced and conjugated to brilliantly colored dyes. Conveniently, ColorBurst Markers require no resuspension, reduction, or heating prior to use.
Colorburst can be used to estimate sample molecular weights, to monitor the progress of an electrophoretic run, or to confirm that an electroblot is complete.

Components

Solution contains myosin-violet, BSA-red, GDH-blue, ADH-red, Carbonic Anhydrase-orange, Trypsin Inhibitor-blue, Lysozyme-red, Aprotinin-blue, 200 mM Tris, SDS and, formamide. Each lot provides the apparent molecular weights determined by comparison to an unconjugated standard.

Quantity

Each vial of ColorBurst contains 500 μL of solution, enough for at least 50 mini gel applications.

Legal Information

ColorBurst is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nidhi Nath et al.
International immunopharmacology, 88, 106925-106925 (2020-09-02)
Expression and single nucleotide polymorphisms (SNPs) of TLR4/9 and CYP1A1 genes are vital for cervical squamous cell carcinoma (CSCC) but considerably vary in different populations. A total of 255-subjects from Jharkhand (130-cases, 125-controls) were utilized to obtain the expression/SNP status
Yi-Shu Huang et al.
International journal of molecular sciences, 22(4) (2021-03-07)
Epidermal growth factor receptor (EGFR) specific therapeutics is of great importance in cancer treatment. Fcy-hEGF fusion protein, composed of yeast cytosine deaminase (Fcy) and human EGF (hEGF), is capable of binding to EGFR and enzymatically convert 5-fluorocytosine (5-FC) to 1000-fold
Meilyn Rodriguez-Hernandez et al.
Protein and peptide letters, 27(2), 145-157 (2019-10-18)
Glycogen storage disease type III (GSDIII, Cori/Forbes disease) is a metabolic disorder due to the deficiency of the Glycogen Debranching Enzyme (GDE), a large monomeric protein (about 176 kDa) with two distinct enzymatic activities: 4-α-glucantransferase and amylo-α-1,6-glucosidase. Several mutations along
Silvia Carloni et al.
Experimental neurology, 324, 113117-113117 (2019-11-18)
Previous studies have shown that simvastatin (Sim) has neuroprotective effects in a neonatal model of hypoxia-ischemia (HI)-induced brain injury when administered before but not after HI, pointing to the preconditioning (PC)-like effects of the statin. The present study aimed to
V A Dzreyan et al.
Molecular neurobiology, 58(1), 217-228 (2020-09-12)
Nerve injury is an important reason of human disability and death. We studied the role of histone deacetylation in the response of the dorsal root ganglion (DRG) cells to sciatic nerve transection. Sciatic nerve transection in the rat thigh induced

Articles

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service