Skip to Content
Merck
All Photos(1)

Key Documents

F6636

SAFC

Nutrient Mixture F-12 Ham

Coon′s Modification, with ʟ-glutamine and zinc sulfate, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

Ham’s F-12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

Nutrient Mixture F-12 Ham, powder, with L-glutamine and 0.863 mg/L zinc sulfate, without sodium bicarbonate, Coon′s Modification, suitable for cell culture

form

powder

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

components

phenol red: 0.00125 g/L
L-glutamine: 0.292 g/L
sodium pyruvate: 0.22 g/L
glucose: 1.802 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Coon's modification of Ham's F-12 was developed for culturing hybrid cells that were produced by viral fusion. The modification consists of doubling the amino acids and pyruvate and including ascorbic acid. The salt concentrations have also been altered. The formula contains 0.863 mg/L zinc sulfate, which may render it unsuitable for culturing mouse L-cells.

Quantity

Formulated to contain 11.5 grams of powder per liter of medium.

Reconstitution

Supplement with 2.68 g/L sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojiao Xue et al.
Human molecular genetics, 26(16), 3116-3129 (2017-06-03)
In-frame premature termination codons (PTCs) account for ∼11% of all disease-associated mutations. PTC suppression therapy utilizes small molecules that suppress translation termination at a PTC to restore synthesis of a full-length protein. PTC suppression is mediated by the base pairing
Kenneth A Giuliano et al.
SLAS discovery : advancing life sciences R & D, 23(2), 111-121 (2017-09-13)
Cystic fibrosis (CF) is a lethal genetic disorder caused by mutation of the cystic fibrosis transmembrane conductance regulator (CFTR) gene. Despite recent groundbreaking approval of genotype-specific small-molecule drugs, a significant portion of CF patients still lack effective therapeutic options that
Feng Liang et al.
SLAS technology, 22(3), 315-324 (2017-04-06)
Cystic fibrosis (CF) is a hereditary disease caused by mutations in the gene coding for the cystic fibrosis transmembrane conductance regulator (CFTR). A large number of nearly 2000 reported mutations, including the premature termination codon (PTC) mutations, urgently require new
Wren E Michaels et al.
Nucleic acids research, 48(13), 7454-7467 (2020-06-11)
Cystic fibrosis (CF) is an autosomal recessive disorder caused by mutations in the CF transmembrane conductance regulator (CFTR) gene, encoding an anion channel that conducts chloride and bicarbonate across epithelial membranes. Mutations that disrupt pre-mRNA splicing occur in >15% of
Nyla Naim et al.
Methods in molecular biology (Clifton, N.J.), 2173, 201-216 (2020-07-12)
cAMP is a crucial mediator of multiple cell signaling pathways. This cyclic nucleotide requires strict spatiotemporal control for effective function. Light-activated proteins have become a powerful tool to study signaling kinetics due to having quick on/off rates and minimal off-target

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Questions

  1. What is the recommended pH of this media?

    1 answer
    1. The pH specification range for this product is 5.4 - 6.0, however this is not suitable as a final pH for the culture of cells. This formulation requires the addition of sodium bicarbonate as a buffering agent. The final pH will be adjusted using an acid or base to reach the desired final pH, typically 7.2 - 7.4. See the links below to review a sample Certificate of Analysis as well as the product datasheet which includes preparation instructions.

      Product datasheet
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/124/814/f6636pis.pdf

      Sample Certificate of Analysis
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/F6636-10X1LSLCP6322.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service