Skip to Content
Merck
All Photos(3)

Key Documents

D8187

Sigma-Aldrich

JumpStart REDTaq® DNA Polymerase

Hot-start Taq enzyme with inert dye, 10X buffer included

Synonym(s):

Hot start DNA polymerase, Hot start Taq

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.55
Pricing and availability is not currently available.

form

liquid

usage

sufficient for 250 reactions
sufficient for 2500 reactions
sufficient for 50 reactions

feature

dNTPs included: no
hotstart

concentration

1 unit/μL

technique(s)

PCR: suitable

color

red

input

purified DNA

suitability

suitable for PCR

shipped in

wet ice

storage temp.

−20°C

General description

JumpStart REDTaq® DNA Polymerase is Sigma′s high performance Taq DNA Polymerase blended with JumpStart Taq antibody and an inert red dye tracer. Extensive testing with a variety of primers and templates indicates that the performance of JumpStart REDTaq DNA Polymerase is equivalent to, or better than, that of standard Taq polymerase.
Since the red tracer has no effect on the amplification process, a sample can be easily re-amplified such as in “nested PCR”. The presence of the dye also has no effect on automated DNA sequencing; ligase mediated ligations, exonucleolytic PCR product digestion, and transformation. Though exceptions may exist, the dye is generally inert in restriction enzyme digestions. If necessary, the dye can be removed from the amplicon by routine purification methodologies.

Application

JumpStart REDTaq® DNA Polymerase has been used in polymerase chain reaction (PCR) for the amplification of:
  • insulin-enterotoxin ricin fusion gene (INS-RTB)[1]
  • endothelial cells DNA derived from reverse transcribed RNA[2]
  • leg genomic DNA from cricket flies[3][4]
  • mitochondrial gene by conventional PCR[5]
JumpStart REDTaq® DNA Polymerase is also suitable for DNA methylation analysis.[6]

Features and Benefits

  • Reduces non-specific amplification
  • Increased target yield and specificity
  • Higher the amplification irrespective of the target concentration
  • Reduce set-up time and eliminate concerns associated with manual or wax hot start methods
  • Visual confirmation that the enzyme has been added and that proper component mixing of the reaction has occurred
  • Samples can be loaded directly onto an agarose gel for electrophoresis without loading buffers or tracking dyes
  • Assembled PCR reactions can be placed at room temperature for up to 2 hours

Packaging

The enzyme is provided with an optimized 10× reaction buffer.

Unit Definition

One unit incorporates 10 nmol of total dNTPs into acid-precipitable DNA in 30 min. at 74 °C.

Other Notes

View more detailed information on JumpStart REDTaq enzymes at www.sigma-aldrich.com/hotstart.

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: US 8,404,464 and US 7,972,828. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims.
JumpStart is a trademark of Sigma-Aldrich Co. LLC
REDTaq is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The field cricket Gryllus assimilis and two new sister species (Orthoptera: Gryllidae)
Weissman DB, et al.
Annals of the Entomological Society of America, 102(3), 367-380 (2009)
The field cricket Gryllus assimilis and two new sister species (Orthoptera: Gryllidae).
Weissman, D.B., et al.
Annals of the Entomological Society of America, 102, 367-367 (2009)
Contributions of VEGF to age-dependent transmural gradients in contractile protein expression in ovine carotid arteries
Butler SM, et al.
American Journal of Physiology. Cell Physiology, 301(3), C653-C666 (2011)
Sarah D Burnett et al.
Journal of toxicology and environmental health. Part A, 84(24), 1020-1039 (2021-08-25)
Inter-species differences in toxicodynamics are often a critical source of uncertainty in safety evaluations and typically dealt with using default adjustment factors. In vitro studies that use cells from different species demonstrated some success for estimating the relationships between life
Expression of a ricin toxin B subunit: insulin fusion protein in edible plant tissues
Carter JE, et al.
Molecular Biotechnology, 44(2), 90-100 (2010)

Articles

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Protocols

Reviews the applications and benefits for RedTaq, including standard RedTaq, Hot Start RedTaq and RedTaq for genomic DNA PCR.

Protocol using antibody mediated hot start polymerase with a red dye for easy gel loading. Method has short activation period (<1min), and results in higher yields and more specificity over standard PCR methods.

Protocol using antibody mediated hot start polymerase. Method has short activation period (<1min), and results in higher yields and more specificity over standard PCR methods.

Protocol using antibody mediated hot start polymerase. Method has short activation period (<1min), and results in higher yields and more specificity over standard PCR methods.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service