Skip to Content
Merck
All Photos(1)

Key Documents

D6063

Sigma-Aldrich

REDTaq® SuperPak DNA Polymerase

Taq for routine PCR with inert dye; with 10X buffer & dNTP mix

Synonym(s):

DNA polymerase with dNTP, DNA polymerase with loading dye, Taq DNA polymerase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

form

liquid

usage

sufficient for 250 reactions

feature

dNTPs included
hotstart: no

concentration

1 unit/μL

technique(s)

PCR: suitable

color

red

input

purified DNA

suitability

suitable for PCR

shipped in

wet ice

storage temp.

−20°C

General description

REDTaq® SuperPak DNA Polymerase is Sigma′s Taq DNA Polymerase mixed with an inert red dye. REDTaq® SuperPak DNA Polymerase is a convenient package, that includes all the necessary components for a PCR reaction except primers, DNA template and water. It consists of Sigma′s high quality REDTaq DNA polymerase, 10 mM ultrapure deoxynucleotide mix and 10´ PCR reaction buffer. The dye provides quick recognition of reactions to which enzyme has been added as well as visual confirmation of complete mixing.

Application

REDTaq® SuperPak DNA Polymerase has been used in:
  • reverse transcriptase-polymerase chain reaction (RT-PCR)
  • nested PCR
  • routine PCR amplification

Features and Benefits

  • Contains inert dye that enables visual confirmation of enzyme addition and direct loading onto agarose gel without the need for an additional loading dye
  • Suitable for nested PCR
  • Contains ultrapure 10mM dNTP mix in a separate vial

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: US 8,404,464 and US 7,972,828. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims.
REDTaq is a registered trademark of Merck KGaA, Darmstadt, Germany
SuperPak is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • P219210X PCR Buffer, Optimized for routine PCR with MgCl2 includedSDS

  • D7295Deoxynucleotide Mix, 10 mM, Molecular Biology ReagentSDS

related product

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Yeste-Velasco et al.
Neuropharmacology, 53(2), 295-307 (2007-07-07)
Recent studies have demonstrated that neuronal reentry in the cell cycle and specifically the expression of the transcription factor E2F-1, constitutes a pathway that may be involved in neuronal apoptosis after serum and potassium withdrawal. Other enzymes such as glycogen
J L Gonzales et al.
Veterinary parasitology, 146(1-2), 9-16 (2007-03-22)
Trypanosomosis caused by Trypanosoma vivax has been a constraint for cattle production in the Bolivian lowlands, since it was introduced in 1996. Flooded areas like the Bolivian Pantanal have a suitable environment for the presence and transmission of Salivarian trypanosomes
A Jóźwik et al.
Veterinary research communications, 29(4), 347-359 (2005-03-09)
Two pairs of primers were prepared, both localized within the sequences of the nucleoprotein gene (NP) of canine distemper virus (CDV). A number of experiments were done to optimize the conditions of RT-PCR and nested PCR methods. The nucleic acids
M Yeste-Velasco et al.
Neuroscience, 159(3), 1135-1147 (2009-04-10)
Increasing evidence implicates the c-Jun NH(2)-terminal kinase (JNK) pathway in the regulation of apoptosis in neurodegenerative diseases. In this study, we examined the neuroprotective effect of SP600125, a selective JNK inhibitor, in cerebellar granule cells (CGNs) deprived of serum and
Innis, M.A., et al.
PCR Protocols: A Guide to Methods and Applications (1990)

Protocols

Reviews the applications and benefits for RedTaq, including standard RedTaq, Hot Start RedTaq and RedTaq for genomic DNA PCR.

Reactions using REDTaq® DNA polymerase are formulated as any PCR mixtures. There are no additional reaction preparation steps or protocol changes required.

Reactions using REDTaq® DNA polymerase are formulated as any PCR mixtures. There are no additional reaction preparation steps or protocol changes required.

Reactions using REDTaq® DNA polymerase are formulated as any PCR mixtures. There are no additional reaction preparation steps or protocol changes required.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service