Skip to Content
Merck
All Photos(1)

Key Documents

T7791

Sigma-Aldrich

Thymidine 5′-triphosphate sodium salt solution

10 mM (pH 7.0)

Synonym(s):

TTP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.52
Pricing and availability is not currently available.

Assay

≥99%

form

liquid

concentration

10 mM (pH 7.0)

color

colorless

foreign activity

DNase, RNase, none detected

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na].CC1=CN(C2CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O2)C(=O)NC1=O

InChI

1S/C10H17N2O14P3.Na.H/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(24-8)4-23-28(19,20)26-29(21,22)25-27(16,17)18;;/h3,6-8,13H,2,4H2,1H3,(H,19,20)(H,21,22)(H,11,14,15)(H2,16,17,18);;

InChI key

KAELIXTVSANDSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thymidine 5′-triphosphate sodium salt solution is used in DNA synthesis reactions such as PCR, DNA sequencing, cDNA synthesis and molecular cloning techniques.

Application

Thymidine 5′-triphosphate sodium salt solution has been used:

  • as a standard to quantify each dNTP from cellular lysates by the liquid chromatography-mass spectrometry (LC-MS/MS) method[1]
  • as a component of the dNTP mix for labeling of ribosomal 18s rDNA probe and telomeric (TTAGG)n probe[2]
  • for routine PCR amplifications

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinhui Chen et al.
Biomedical chromatography : BMC, 31(3) (2016-08-25)
The endogenous deoxynucleoside triphosphate (dNTP) pool includes deoxyadenosine triphosphate (dATP), deoxycytidine triphosphate (dCTP), deoxyguanosine triphosphate (dGTP) and thymidine triphosphate (TTP). The endogenous dNTP pool is regulated by complex enzymatic pathways that can be targeted by drugs, such as antimetabolites. In
Michael G Mohsen et al.
Nucleic acids research, 47(18), 9495-9501 (2019-09-11)
We document the preparation and properties of dimerized pentaphosphate-bridged deoxynucleotides (dicaptides) that contain reactive components of two different nucleotides simultaneously. Importantly, dicaptides are found to be considerably more stable to hydrolysis than standard dNTPs. Steady-state kinetics studies show that the

Protocols

Protocol using antibody mediated hot start polymerase with a red dye for easy gel loading. Method has short activation period (<1min), and results in higher yields and more specificity over standard PCR methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service