Skip to Content
Merck
All Photos(2)

Key Documents

P2893

Sigma-Aldrich

JumpStart Taq ReadyMix

Complete optimized reagent for hot-start PCR at 2X concentration

Synonym(s):

hot start PCR master mix, hot start master mix, hot start taq master mix, mutiplex PCR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106300
NACRES:
NA.55

form

liquid

usage

sufficient for 100 reactions
sufficient for 400 reactions

feature

Multiplex PCR
dNTPs included
hotstart

concentration

2.5 units/reaction (50 μL reaction volume)

technique(s)

PCR: suitable

color

colorless

suitability

suitable for (quantitative PCR)

application(s)

agriculture

shipped in

wet ice

storage temp.

−20°C

General description

JumpStart Taq ReadyMix is a ready-to-use 2X master mix that contains JumpStart Taq DNA polymerase, 99% pure dNTPs, reaction buffer and JumpStart Taq antibody. The Taq DNA polymerase is an antibody-inactivated hot-start enzyme. Once the reaction temperature reaches 70°C, the DNA polymerase-antibody complex dissociates and Taq DNA polymerase activity is restored. This antibody-enzyme complex allows for easy and convenient set-up with less contamination risk than manual hot-start techniques.

Application

JumpStart Taq ReadyMix has been used in quantitative polymerase chain reaction (PCR). It has also been used to identify fibroblast cell surface adenosine receptor (AR) types under hypoxia. It is also suitable:
  • For PCR amplifications that require reduced non-specific amplification
  • For multiplex PCR
  • For reduction of primer dimers

Features and Benefits

  • The master mix allows consistency from one reaction to the next
  • Reduced preparation time and reduced risk of contamination from multiple pipetting steps
  • Designed to minimize non-specific amplification and contamination
  • Increased specificity and target yield
  • Reduced primer dimers
  • Reduced set-up time as compared to manual or wax Hot Start methods
  • Allows for room temperature set-up
  • Ideal for high throughput, quantitative PCR applications

Packaging

Default reaction volume is 50 μL

100RXN is packaged as 1 X 2.5 mL
400RXN is packaged as 1 X 10 mL

Other Notes

JumpStart Taq ReadyMix is a prepared solution combining the performance benefits of hot start PCR with the convenience of a ReadyMix. The mix includes JumpStart Taq DNA polymerase, 99% pure deoxynucleotides and buffer in a 2× optimized reaction concentrate. JumpStart Taq Polymerase is an antibody-inactivated hot-start enzyme designed to minimize non-specific amplification while increasing target yield. Unlike other hot-start methods (i.e. chemical inactivation), JumpStart Taq polymerase does not require a pre-incubation step prior to cycling because polymerase activity is fully restored during the first denaturation cycle of the PCR reaction. The hot start mechanism allows for room temperature set up, making it ideal for high throughput applications. To prepare a 50 μL reaction, simply add 25 μL of ReadyMix to template, primers and water for a final reaction volume of 50 μL.
To prepare a 50 μL reaction, simply add 25 μL of ReadyMix to template, primers and water for a final reaction volume of 50 μL.
This product has been validated in quantitative PCR, but may require supplementation with magnesium chloride solution, 25 mM, Catalog Number M8787, a suitable fluorescent probe, and, if desired, an internal reference dye, Catalog Number R4526. View more detailed information on JumpStart Taq ReadyMix products at www.sigma-aldrich.com/hotstart.

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: US 8,404,464 and US 7,972,828. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims.
JumpStart is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of A2BR on the biological behavior of mouse renal fibroblasts during hypoxia
Tang Jin, et al.
Molecular Medicine Reports, 11(6), 4397-4402 (2015)
Koen Kole et al.
GigaScience, 6(10), 1-6 (2017-10-13)
Experience-dependent plasticity (EDP) is essential for anatomical and functional maturation of sensory circuits during development. Although the principal synaptic and circuit mechanisms of EDP are increasingly well studied experimentally and computationally, its molecular mechanisms remain largely elusive. EDP can be
Madhavi A Naik et al.
International journal of surgical pathology, 28(4), 371-381 (2019-12-25)
Cystic neutrophilic granulomatous mastitis (CNGM) is a histologically characterized variant of granulomatous lobular mastitis that is associated with lipophilic Corynebacterium species. It remains a largely underrecognized entity in India. Our aim was to study CNGM in the Asian Indian population
Claudia Eberle et al.
PloS one, 7(9), e45361-e45361 (2012-10-03)
Maternal immunization with oxidized lipoproteins prior to pregnancy protects against atherogenic in utero programming by gestational hypercholesterolemia and enhances beneficial lymphocyte-dependent immune responses in offspring. To determine whether in utero programming and immunomodulation also affect insulin resistance (IR) and type
Bini Mathew et al.
European journal of medicinal chemistry, 210, 112952-112952 (2020-11-04)
ALS is a rare type of progressive neurological disease with unknown etiology. It results in the gradual degeneration and death of motor neurons responsible for controlling the voluntary muscles. Identification of mutations in the superoxide dismutase (SOD) 1 gene has

Articles

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

See All

Protocols

Protocol using antibody mediated hot start polymerase. Method has short activation period (<1min), and results in higher yields and more specificity over standard PCR methods.

Protocol using antibody mediated hot start polymerase. Method has short activation period (<1min), and results in higher yields and more specificity over standard PCR methods.

Protocol using antibody mediated hot start polymerase. Method has short activation period (<1min), and results in higher yields and more specificity over standard PCR methods.

Related Content

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service