Skip to Content
Merck
All Photos(1)

Key Documents

D6665

Sigma-Aldrich

2′,7′-Dichlorofluorescein

~90% purity (TLC) (85- 100% purity), powder

Synonym(s):

Dichlorofluorescein; Fluorescein 27

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Cl2O5
CAS Number:
Molecular Weight:
401.20
Beilstein:
58009
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

2′,7′-Dichlorofluorescein, suitable for use as an indicator in chloride titration, ~90% (TLC), powder

Assay

~90% (TLC)

form

powder

technique(s)

titration: suitable

mp

280 °C (dec.) (lit.)

solubility

ethanol: 25 mg/mL

density

0.79 g/cm3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2Oc3cc(O)c(Cl)cc3C4(OC(=O)c5ccccc45)c2cc1Cl

InChI

1S/C20H10Cl2O5/c21-13-5-11-17(7-15(13)23)26-18-8-16(24)14(22)6-12(18)20(11)10-4-2-1-3-9(10)19(25)27-20/h1-8,23-24H

InChI key

VFNKZQNIXUFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′,7′-Dichlorofluorescein is an oxidation-sensitive fluorescent probe. 2′7′-dichlorofluorescein diacetate is oxidized to 2′7′-dichlorofluorescein in the presence of ROS (reactive oxygen species).citation

Application

2′,7′-Dichlorofluorescein has been used as an oxidation-sensitive fluorescent probe to measure ROS (reactive oxygen species) formation in cells.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vittoria Buccigrossi et al.
PloS one, 9(6), e99830-e99830 (2014-06-12)
Rotavirus (RV) infection causes watery diarrhea via multiple mechanisms, primarily chloride secretion in intestinal epithelial cell. The chloride secretion largely depends on non-structural protein 4 (NSP4) enterotoxic activity in human enterocytes through mechanisms that have not been defined. Redox imbalance
Cysteamine suppresses human peripheral blood mononuclear cells--human corneal endothelial cell reaction via reactive oxygen species reduction.
Shin YJ, et al.
Molecular Vision, 17, 3371-3378 (2011)
Olga N Pakhomova et al.
Archives of biochemistry and biophysics, 527(1), 55-64 (2012-08-23)
Nanosecond pulsed electric field (nsPEF) is a novel modality for permeabilization of membranous structures and intracellular delivery of xenobiotics. We hypothesized that oxidative effects of nsPEF could be a separate primary mechanism responsible for bioeffects. ROS production in cultured cells
M-C Chang et al.
Journal of periodontal research, 48(1), 66-73 (2012-07-28)
Short-chain fatty acids, such as butyric acid and propionic acid, are metabolic by-products generated by periodontal microflora such as Porphyromonas gingivalis, and contribute to the pathogenesis of periodontitis. However, the effects of butyrate on the biological activities of gingival fibroblasts
Andréa G K Ferreira et al.
Journal of cellular biochemistry, 113(1), 174-183 (2011-09-02)
The present study investigated the effects of chronic hyperprolinemia on oxidative and metabolic status in liver and serum of rats. Wistar rats received daily subcutaneous injections of proline from their 6th to 28th day of life. Twelve hours after the

Questions

1–4 of 4 Questions  
  1. What excitation /emission wavelengths are best for fluorescence detection of 2´,7´-Dichlorofluorescein?

    1 answer
    1. 2´,7´-Dichlorofluorescein can be detected at an excitation wavelength at 504 nm and emission wavelength at 529 nm in 0.1 M Tris pH 8.0.

      Helpful?

  2. What can be used to make a solution of 2´,7´-dichlorofluorescein?

    1 answer
    1. 2´,7´-Dichlorofluorescein is soluble at 25 mg/mL absolute ethanol with heating. 2´,7´-Dichlorofluorescein is also soluble in water at 0.1 mg/mL.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. In what types of applications can 2´,7´-Dichlorofluorescein be used?

    1 answer
    1. Some applications for use of 2´,7´-Dichlorofluorescein are listed below. This product can be used for the determination of carbohydrates by fluorescence densitometry after TLC.  See G. Gübitz et al., J. Chromatogr., 117, 337, (1976). 2´,7´-Dichlorofluorescein as also been used in the determination of H2O2 by chemiluminescence. See the following references by H. Watanabe et al. Anal. Sci. 2, 461, (1986) and T. Segawa et al. Anal. Sci. 6, 763, (1990). This product has also been used as a  fluorescent additive used in indirect fluorimetric detection in OTC-HPLC. See W.D. Pfeffer and E.S. Yeung, J. Chromatogr., 506, 401, (1990).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service